Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Iodonitrotetrazolium chloride
Synonym(s):
2-(4-Iodophenyl)-3-(4-nitrophenyl)-5-phenyl-2
H
-tetrazolium chloride,
p
-Iodonitrotetrazolium Violet, INT
Empirical Formula (Hill Notation):
C
19
H
13
ClIN
5
O
2
CAS Number:
146-68-9
Molecular Weight:
505.70
EC Number:
205-676-2
Products
Product Category
building blocks (1)
stains (1)
Brand
Sigma-Aldrich (3)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Markush Class
Markush Group
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
I10406
Iodonitrotetrazolium chloride,
95%
Expand
View Pricing
I8377
Iodonitrotetrazolium chloride,
Used in colorimetric assays.
Expand
View Pricing
58030
Iodonitrotetrazolium chloride,
BioReagent, ≥97.0% (calc. on dry substance, NT)
Expand
View Pricing