Skip to Content
Merck
All Photos(1)

Key Documents

07-5990

Sigma-Aldrich

2,2′-Dipyridyl

JIS special grade, ≥99.0%

Synonym(s):

2,2′-Bipyridyl, 2,2′-Bipyridine, 2,2′-Dipyridine, 2,2′-Dipyridyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8N2
CAS Number:
Molecular Weight:
156.18
Beilstein:
113089
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

JIS special grade

Assay

≥99.0%

availability

available only in Japan

bp

273 °C (lit.)

mp

70-73 °C (lit.)

SMILES string

c1ccc(nc1)-c2ccccn2

InChI

1S/C10H8N2/c1-3-7-11-9(5-1)10-6-2-4-8-12-10/h1-8H

InChI key

ROFVEXUMMXZLPA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Metalloprotease inhibitor, high-affinity chelator of iron; may inhibit Fe2+ containing enzymes at 10−8 M.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

267.8 °F - closed cup

Flash Point(C)

131 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metal-locked DNA three-way junction.
Jean-Louis H A Duprey et al.
Angewandte Chemie (International ed. in English), 52(4), 1212-1216 (2012-12-15)
Ralf Haiges et al.
Angewandte Chemie (International ed. in English), 54(31), 9101-9105 (2015-06-16)
Vanadium(V) oxoazide [VO(N3)3] was prepared through a fluoride-azide exchange reaction between [VOF3] and Me3SiN3 in acetonitrile solution. When the highly impact- and friction-sensitive compound [VO(N3)3] was reacted with 2,2′-bipyridine, the adduct [(bipy)VO(N3)3] was isolated. The reaction of [VO(N3)3] with [PPh4]N3
Hana Krysova et al.
Chimia, 67(3), 149-154 (2013-04-12)
Titania nanofibers were fabricated using the industrial Nanospider(TM) technology. The preparative protocol was optimized by screening various precursor materials to get pure anatase nanofibers. Composite films were prepared by mixing a commercial paste of nanocrystalline anatase particles with the electrospun
Yanyan Wang et al.
Langmuir : the ACS journal of surfaces and colloids, 29(4), 1273-1279 (2013-01-05)
In this paper, the fabrication, characterization, and application in oxygen sensing are reported for a novel multifunctional nanomaterial of [Ru(bpy)(2)phen-MMS] (bpy, 2,2'-bipyridyl; phen, phenathrolin) which was simply prepared by covalently grafting the ruthenium(II) polypyridyl compounds into the channels of magnetic
Xiang Chen et al.
Dalton transactions (Cambridge, England : 2003), 42(13), 4386-4397 (2013-02-13)
Two ruthenium(II) polypyridyl complexes, [Ru(bpy)(2)(ptpn)](2+) (1) (bpy = 2,2'-bipyridine, ptpn = 3-(1,10-phenanthroline-2-yl)-as-triazino[5,6-f]1,10-phenanthroline) and [Ru(phen)(2)(ptpn)](2+) (2) (phen = 1,10-phenanthroline), were synthesized and characterized. Crystal structure analysis shows that complex 1 has a large planar aromatic area and possesses the potential to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service