Skip to Content
Merck
All Photos(2)

Key Documents

754730

Sigma-Aldrich

Tris(2,2′-bipyridine)ruthenium(II) hexafluorophosphate

97%

Synonym(s):

Ru(bpy)3(PF6)2, Ruthenium-tris(2,2′-bipyridyl) dihexafluorophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H24F12N6P2Ru
CAS Number:
Molecular Weight:
859.55
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

solid

reaction suitability

core: ruthenium
reaction type: Photocatalysis
reagent type: catalyst

impurities

(Material is not fluorescence active.)

mp

>300 °C

photocatalyst activation

425 nm

absorption

13,400 at 456 nm
80,000 at 291 nm

SMILES string

[Ru++].F[P-](F)(F)(F)(F)F.F[P-](F)(F)(F)(F)F.c1ccc(nc1)-c2ccccn2.c3ccc(nc3)-c4ccccn4.c5ccc(nc5)-c6ccccn6

InChI

1S/3C10H8N2.2F6P.Ru/c3*1-3-7-11-9(5-1)10-6-2-4-8-12-10;2*1-7(2,3,4,5)6;/h3*1-8H;;;/q;;;2*-1;+2

InChI key

KLDYQWXVZLHTKT-UHFFFAOYSA-N

General description

Tris(2,2′-bipyridine)ruthenium(II) hexafluorophosphate (Ru(bpy)3(PF6)2) is a conducting polymer that is majorly used as an active layer in electrochemical devices. It facilitates the formation of highly efficient and low voltage devices. It shows an external quantum efficiency of 0.35 and 400 cd/m2 at 3V.

Application

Ru(bpy)3(PF6)2 may be used as a conjugating polymer in the development of light emitting electrochemical cell based devices such as light emitting diodes (LEDs). It is also used as a high-efficiency triplet emitter for OLED/Sensor research.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wesley R Browne et al.
Journal of the American Chemical Society, 125(7), 1706-1707 (2003-02-13)
The excited-state properties of the transition metal complexes tris(2,2'-bipyridine) ruthenium(II) and tris(4,7-diphenyl-1,10-phenanthroline) ruthenium(II) are examined using picosecond time-resolved luminescence spectroscopy. For both complexes, direct observation of a short-lived high-energy emission with a lifetime of less than 4 ps is reported.
Silvia Voci et al.
Analytical and bioanalytical chemistry, 412(17), 4067-4075 (2020-04-29)
Nanofluidic electrochemical devices confine the volume of chemical reactions to femtoliters. When employed for light generation by electrochemiluminescence (ECL), nanofluidic confinement yields enhanced intensity and robust luminescence. Here, we investigate different ECL pathways, namely coreactant and annihilation ECL in a
Valentina Leandri et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 20(4), 618-626 (2019-01-10)
The excited state dynamics of Tris(2,2'-bipyridine)ruthenium(II) hexafluorophosphate, [Ru(bpy)3 (PF6 )2 ], was investigated on the surface of bare and sensitized TiO2 and ZrO2 films. The organic dyes LEG4 and MKA253 were selected as sensitizers. A Stern-Volmer plot of LEG4-sensitized TiO2
Direct measurement of the electric-field distribution in a light-emitting electrochemical cell
Slinker JD, et al.
Testing, 6(11), 894-894 (2007)
Thin-film solid-state electroluminescent devices based on tris (2, 2 `-bipyridine) ruthenium (II) complexes
Buda, M
Journal of the American Chemical Society, 124(21), 6090-6098 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service