Skip to Content
Merck
All Photos(1)

Key Documents

C2460000

Clozapine

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

8-Chloro-11-(4-methyl-1-piperazinyl)-5H-dibenzo[b,e][1,4]-diazepine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H19ClN4
CAS Number:
Molecular Weight:
326.82
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

clozapine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CN1CCN(CC1)C2=Nc3cc(Cl)ccc3Nc4ccccc24

InChI

1S/C18H19ClN4/c1-22-8-10-23(11-9-22)18-14-4-2-3-5-15(14)20-16-7-6-13(19)12-17(16)21-18/h2-7,12,20H,8-11H2,1H3

InChI key

QZUDBNBUXVUHMW-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Clozapine EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bianca A Silva et al.
Nature neuroscience, 16(12), 1731-1733 (2013-11-12)
The neural circuits mediating fear to naturalistic threats are poorly understood. We found that functionally independent populations of neurons in the ventromedial hypothalamus (VMH), a region that has been implicated in feeding, sex and aggression, are essential for predator and
A Merenlender-Wagner et al.
Molecular psychiatry, 20(1), 126-132 (2013-12-25)
Autophagy is a process preserving the balance between synthesis, degradation and recycling of cellular components and is therefore essential for neuronal survival and function. Several key proteins govern the autophagy pathway including beclin1 and microtubule associated protein 1 light chain
Rasmus Røge et al.
Schizophrenia research, 140(1-3), 204-213 (2012-07-27)
Clozapine remains the drug of choice for treatment resistant schizophrenia, but is associated with potentially life threatening side effects, including agranulocytosis and myocarditis. Immunological mechanisms may be involved in the development of these side effects or in the unique antipsychotic
Gary Remington et al.
Psychopharmacology, 225(3), 505-518 (2012-11-28)
Clozapine levels are advocated in the monitoring of patients on this drug and have now been used for a number of years. A safety-related threshold has also been proposed, as well as therapeutic lower and upper thresholds. While there has
Roland Bock et al.
Nature neuroscience, 16(5), 632-638 (2013-04-02)
A hallmark of addiction is the loss of control over drug intake, which is seen in only a fraction of those exposed to stimulant drugs such as cocaine. The cellular mechanisms underlying vulnerability or resistance to compulsive drug use remain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service