Skip to Content
Merck
All Photos(1)

Key Documents

Y0000758

Clozapine for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Clozapine, 8-Chloro-11-(4-methyl-1-piperazinyl)-5H-dibenzo[b,e][1,4]-diazepine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H19ClN4
CAS Number:
Molecular Weight:
326.82
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

clozapine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CN1CCN(CC1)C2=Nc3cc(Cl)ccc3Nc4ccccc24

InChI

1S/C18H19ClN4/c1-22-8-10-23(11-9-22)18-14-4-2-3-5-15(14)20-16-7-6-13(19)12-17(16)21-18/h2-7,12,20H,8-11H2,1H3

InChI key

QZUDBNBUXVUHMW-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Clozapine for peak identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bianca A Silva et al.
Nature neuroscience, 16(12), 1731-1733 (2013-11-12)
The neural circuits mediating fear to naturalistic threats are poorly understood. We found that functionally independent populations of neurons in the ventromedial hypothalamus (VMH), a region that has been implicated in feeding, sex and aggression, are essential for predator and
Gary Remington et al.
Psychopharmacology, 225(3), 505-518 (2012-11-28)
Clozapine levels are advocated in the monitoring of patients on this drug and have now been used for a number of years. A safety-related threshold has also been proposed, as well as therapeutic lower and upper thresholds. While there has
Chun-Hsin Chen et al.
The Journal of clinical psychiatry, 74(5), e424-e430 (2013-06-14)
Many studies have shown that metformin can decrease body weight and improve metabolic abnormalities in patients with schizophrenia. Whether or not the beneficial effects can be sustained after discontinuation of metformin needs to be evaluated. We conducted a 24-week randomized
Shalini Jain et al.
The Journal of clinical investigation, 123(4), 1750-1762 (2013-03-13)
Type 2 diabetes (T2D) has emerged as a major threat to human health in most parts of the world. Therapeutic strategies aimed at improving pancreatic β cell function are predicted to prove beneficial for the treatment of T2D. In the
A Merenlender-Wagner et al.
Molecular psychiatry, 20(1), 126-132 (2013-12-25)
Autophagy is a process preserving the balance between synthesis, degradation and recycling of cellular components and is therefore essential for neuronal survival and function. Several key proteins govern the autophagy pathway including beclin1 and microtubule associated protein 1 light chain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service