Skip to Content
Merck
All Photos(1)

Key Documents

96109

Supelco

Anisole

analytical standard

Synonym(s):

Methoxybenzene, Methyl phenyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H5
CAS Number:
Molecular Weight:
108.14
Beilstein:
506892
EC Number:
MDL number:
UNSPSC Code:
41116107
eCl@ss:
39023603
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

3.7 (vs air)

vapor pressure

10 mmHg ( 42.2 °C)

Assay

≥99.9% (GC)

autoignition temp.

887 °F

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.516 (lit.)
n20/D 1.518

bp

154 °C (lit.)

mp

−37 °C (lit.)

density

0.995 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

COc1ccccc1

InChI

1S/C7H8O/c1-8-7-5-3-2-4-6-7/h2-6H,1H3

InChI key

RDOXTESZEPMUJZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Anisole is a colorless liquid , which can be obtained upon heating anisic acid with an excess of barium hydroxide.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

109.4 °F - closed cup

Flash Point(C)

43 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elements of Chemistry: Including the History of the Imponderables and the Inorganic Chemistry (1846)
Practical Organic Chemistry (1975)
S Rivera et al.
Analytical and bioanalytical chemistry, 400(5), 1339-1346 (2011-03-08)
Various carotenoids were analyzed by ultra-high-pressure liquid chromatography with tandem mass spectrometry detection (UHPLC-MS/MS). Three different techniques to ionize the carotenoids were compared: electrospray ionization (ESI), atmospheric pressure chemical ionization (APCI) and atmospheric pressure photoionization (APPI). For all of the
Laszlo Hollosi et al.
Journal of chromatography. A, 1218(1), 23-31 (2010-12-03)
European food legislation defines a set of 16 polycyclic aromatic hydrocarbons (PAHs) as of high concern for human health. The EU set contains structurally very similar PAHs with ring numbers between 4 and 6, and so raises some separation aspects
Kohji Omata et al.
Journal of combinatorial chemistry, 12(4), 435-444 (2010-05-22)
To study the synergistic effect on the acidity and activity of Si-Al-Zr ternary oxide system, the mixture design of experiments was applied to prepare the ternary oxides by a sol-gel method, and rapid screening on dehydration of isopropanol and Friedel-Crafts

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service