Skip to Content
Merck
All Photos(7)

Documents

H9003

Sigma-Aldrich

Hydroquinone

ReagentPlus®, ≥99%

Synonym(s):

1,4-Benzenediol, 1,4-Dihydroxybenzene, HQ

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,4-(OH)2
CAS Number:
Molecular Weight:
110.11
Beilstein:
605970
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic

Quality Level

vapor density

3.81 (vs air)

vapor pressure

1 mmHg ( 132 °C)

product line

ReagentPlus®

Assay

≥99%

form

powder

autoignition temp.

930 °F

bp

285 °C (lit.)

mp

172-175 °C (lit.)

solubility

water: 50 mg/mL, clear

SMILES string

Oc1ccc(O)cc1

InChI

1S/C6H6O2/c7-5-1-2-6(8)4-3-5/h1-4,7-8H

InChI key

QIGBRXMKCJKVMJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hydroquinone is a phenolic compound, which is naturally found in coffee, tea and wine. It is used in photographic, chemical and cosmetic industries. Hydroquinone is used as a skin lightening agent. It is topically applied to treat melisma, freckles and senile lentigines. Hydroquinone is linked to exogenous ochronosis, dermatitis and skin irritation. It inhibits melanogenesis.

Application

Hydroquinone has been used as reagent for the conversion of DNA by sodium bisulfite. It has also been used as a chemical in Timm′s staining.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Muta. 2 - Skin Sens. 1B

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

329.0 °F - closed cup

Flash Point(C)

165 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Xenograft of human umbilical mesenchymal stem cells from Wharton?s jelly as a potential therapy for rat pilocarpine-induced epilepsy
Huang PY, et al.
Brain, behavior, and immunity, 54, 45-58 (2016)
Handbook of Cosmetic Science and Technology (2005)
Procedures in Cosmetic Dermatology Series: Cosmeceuticals E-Book (2008)
Increased frequency of CpG island methylator phenotype and CDH1 methylation in a gastric cancer high-risk region of china
Zhang KL, et al.
Translational Oncology, 1(1), 28-35 (2008)
CDH1 promoter methylation correlates with decreased gene expression and poor prognosis in patients with breast cancer
Liu J, et al.
Oncology Letters, 11(4), 2635-2643 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service