Skip to Content
Merck
All Photos(3)

Key Documents

296295

Sigma-Aldrich

Anisole

anhydrous, 99.7%

Synonym(s):

Methoxybenzene, Methyl phenyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H5
CAS Number:
Molecular Weight:
108.14
Beilstein:
506892
EC Number:
MDL number:
UNSPSC Code:
12352112
eCl@ss:
39023603
PubChem Substance ID:
NACRES:
NA.21
grade:
anhydrous
Assay:
99.7%
bp:
154 °C (lit.)
vapor pressure:
10 mmHg ( 42.2 °C)

grade

anhydrous

Quality Level

vapor density

3.7 (vs air)

vapor pressure

10 mmHg ( 42.2 °C)

Assay

99.7%

form

liquid

autoignition temp.

887 °F

expl. lim.

8 %

impurities

<0.002% water
<0.005% water (100 mL pkg)

evapn. residue

<0.0003%

refractive index

n20/D 1.516 (lit.)

bp

154 °C (lit.)

mp

−37 °C (lit.)

density

0.995 g/mL at 25 °C (lit.)

SMILES string

COc1ccccc1

InChI

1S/C7H8O/c1-8-7-5-3-2-4-6-7/h2-6H,1H3

InChI key

RDOXTESZEPMUJZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Anisole may undergo:
  • Zeolite-catalyzed Friedel-Crafts acylation with acetic anhydride in acetic acid to form 4-methoxyacetophenone with high selectivity.
  • Bismuth triflate-catalyzed Friedel-Crafts benzoylation with benzoic anhydride to form 4-methoxybenzophenone under solvent-free microwave irradiation.

It may also be used as a surrogate for lignin primary tar in pyrolysis experiments.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

125.1 °F - closed cup

Flash Point(C)

51.7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Greener Friedel?Crafts Acylation Using Microwave?Enhanced Reactivity of Bismuth Triflate in the Friedel?Crafts Benzoylation of Aromatic Compounds with Benzoic Anhydride
Tran PH, et al
ChemistrySelect, 2(1), 571-575 (2017)
Selective vapor-phase hydrodeoxygenation of anisole to benzene on molybdenum carbide catalysts.
Lee WS, et al.
J. Catal., 319, 44-53 (2014)
C13 Nuclear magnetic resonance spectroscopy. II. Phenols, anisole and dimethoxybenzenes.
Lauterbur PC.
Journal of the American Chemical Society, 83(8), 1846-1852 (1961)
A Novel Friedel-Crafts Acylation Reaction of Anisole for Production of 4-Methoxyacetophenone with High Selectivity and Sufficient Reusability of Mordenite Zeolite Catalyst
Makihara M & Komura K
Green and Sustainable Chemistry, 7(03), 185-185 (2017)
Vibrational spectra and torsional barriers of anisole and some monohalogen derivatives.
Owen NL and Hester RE.
Spectrochimica Acta Part A: Molecular Spectroscopy, 25(2), 343-354 (1969)

Articles

Fluorescence quenching microscopy visualizes 2D materials like graphene and MoS2 rapidly, inexpensively, and with high fidelity.

Fluorescence quenching microscopy visualizes 2D materials like graphene and MoS2 rapidly, inexpensively, and with high fidelity.

Fluorescence quenching microscopy visualizes 2D materials like graphene and MoS2 rapidly, inexpensively, and with high fidelity.

Fluorescence quenching microscopy visualizes 2D materials like graphene and MoS2 rapidly, inexpensively, and with high fidelity.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service