Skip to Content
Merck
All Photos(1)

Key Documents

74280

Sigma-Aldrich

1-Nonanol

purum, ≥98.0% (GC)

Synonym(s):

Alcohol C9, Nonyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)8OH
CAS Number:
Molecular Weight:
144.25
Beilstein:
969213
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

5 (vs air)

Quality Level

vapor pressure

13 mmHg ( 104 °C)

grade

purum

Assay

≥98.0% (GC)

form

liquid

refractive index

n20/D 1.433 (lit.)
n20/D 1.434

bp

215 °C (lit.)

mp

−8-−6 °C (lit.)

density

0.827 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

CCCCCCCCCO

InChI

1S/C9H20O/c1-2-3-4-5-6-7-8-9-10/h10H,2-9H2,1H3

InChI key

ZWRUINPWMLAQRD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Mougabure Cueto et al.
Journal of medical entomology, 39(3), 457-460 (2002-06-14)
The effectiveness of 1-octanol, 1-nonanol, 1-decanol, 1-undecanol, and 1-dodecanol was evaluated by immersion method against susceptible and permethrin-resistant head lice, Pediculus humanus capitis De Geer, from Buenos Aires, Argentina. All the tested alcohols showed knockdown effect at 10 min and
Mathias Ditzen et al.
Chemical senses, 28(9), 781-789 (2003-12-05)
The brain's link between perception and action involves several steps, which include stimulus transduction, neuronal coding of the stimulus, comparison to a memory template and choice of an appropriate behavioral response. All of these need time, and many studies report
Guanghui Zhao et al.
Journal of hazardous materials, 175(1-3), 715-725 (2009-11-21)
This study investigated the potential of capsules containing 1-nonanol for the adsorption of phenol at high initial concentrations. The polysulfone capsules containing 1-nonanol (PSF@1-nonanol capsules) were successfully prepared with a phase inversion method, and the results showed that 1-nonanol was
Emmanuel Perisse et al.
BMC biology, 7, 30-30 (2009-06-18)
Synaptic plasticity associated with an important wave of gene transcription and protein synthesis underlies long-term memory processes. Calcium (Ca2+) plays an important role in a variety of neuronal functions and indirect evidence suggests that it may be involved in synaptic
D M Greene-McDowelle et al.
Toxicon : official journal of the International Society on Toxinology, 37(6), 883-893 (1999-05-26)
The fungi Aspergillus flavus and Aspergillus parasiticus produce the hepatocarcinogenic, secondary metabolites, aflatoxins, in cottonseed, corn, peanuts and treenuts. Results have shown that aflatoxigenic strains of A. flavus and A. parasiticus grown in the presence of specific cotton-leaf volatiles exhibit

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service