Skip to Content
Merck
All Photos(2)

Key Documents

53877

Supelco

2,6-Dimethoxyphenol

analytical standard

Synonym(s):

Pyrogallol 1,3-dimethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2C6H3OH
CAS Number:
Molecular Weight:
154.16
Beilstein:
1526871
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

261 °C (lit.)

mp

50-57 °C (lit.)
53-57 °C

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

COc1cccc(OC)c1O

InChI

1S/C8H10O3/c1-10-6-4-3-5-7(11-2)8(6)9/h3-5,9H,1-2H3

InChI key

KLIDCXVFHGNTTM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,6-Dimethoxyphenol can find applications mainly as a substrate of laccases catalysis and also determine its activity in aqueous solution. It can also be used as a potential marker compound in particular for emission of smoke in the atmosphere from wood.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

284.0 °F - closed cup

Flash Point(C)

140 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Enzymatic catalysis of 2, 6-dimethoxyphenol by laccases and products characterization in organic solutions
Yang YW, et al.
Science in China. Series B, Chemistry, Life Sciences & Earth Sciences, 51, 669-676 (2008)
Rate constant and secondary organic aerosol yields for the gas-phase reaction of hydroxyl radicals with syringol (2, 6-dimethoxyphenol)
Lauraguais A, et al.
Atmospheric Environment, 55, 43-48 (2013)
Isabel Pardo et al.
Biotechnology and bioengineering, 109(12), 2978-2986 (2012-06-26)
DNA recombination methods are useful tools to generate diversity in directed evolution protein engineering studies. We have designed an array of chimeric laccases with high-redox potential by in vitro and in vivo DNA recombination of two fungal laccases (from Pycnoporus
Jaromir Michałowicz et al.
Chemosphere, 66(4), 657-663 (2006-09-12)
Germs, xenobiotics and organic matter that influence the colour, turbidity and organoloeptic properties of water are removed by chlorination. Unfortunately, chlorine oxidants including sodium hypochlorite, used in water treatment induce processes that partly convert the treated compounds to unwanted chlorinated
Rosario Díaz et al.
Applied microbiology and biotechnology, 88(1), 133-142 (2010-07-08)
Two laccase isoenzymes were purified and characterized from the basidiomycete Coriolopsis rigida during transformation of the water-soluble fraction of "alpeorujo" (WSFA), a solid residue derived from the olive oil production containing high levels of toxic compounds. Zymogram assays of laccases

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service