Skip to Content
Merck
All Photos(1)

Key Documents

D8885

Sigma-Aldrich

Diethanolamine

reagent grade, ≥98.0%

Synonym(s):

2,2′-Iminodiethanol, Bis(2-hydroxyethyl)amine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HN(CH2CH2OH)2
CAS Number:
Molecular Weight:
105.14
Beilstein:
605315
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

grade

reagent grade

Quality Level

vapor density

3.6 (vs air)

vapor pressure

<0.98 atm ( 100 °C)

Assay

≥98.0%

form

solid or viscous liquid

autoignition temp.

689 °F

expl. lim.

10.6 %

color

clear

refractive index

n20/D 1.477 (lit.)

pKa (25 °C)

8.88

bp

217 °C/150 mmHg (lit.)

mp

28 °C (lit.)

density

1.097 g/mL at 25 °C (lit.)

functional group

amine
hydroxyl

SMILES string

OCCNCCO

InChI

1S/C4H11NO2/c6-3-1-5-2-4-7/h5-7H,1-4H2

InChI key

ZBCBWPMODOFKDW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diethanolamine (DEA) is an amino alcohol commonly used in the preparation of soaps and surfactants, agricultural chemicals and in textile processing. It is used as an absorbent for capturing CO2. Its toxic and carcinogenic effect has been studied.

Application

Diethanolamine has been used:
  • As a solvent and stabilizer to synthesize zinc oxide (ZnO): indium (In) thin film, which was employed as photoanode in dye synthesized solar cell (DSSC).
  • To prepare diethanolamine substrate buffer.
  • To prepare a pH10 buffer solution along with MgCl2 for the incubation of cell lysates extracted from astrocyte cultures.
It may be used in the following processes:
  • Preparation of disodiumiminodiacetic acid by dehydrogenation.
  • As a solvent and reactant in triazine based dendrimer synthesis.
  • Preparation of diethanolamine ionic liquids employed in green Heck reaction.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Oral

Target Organs

Kidney,Liver,Blood

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

280.4 °F - closed cup

Flash Point(C)

138 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Fabrication and characterization of ZnO: In thin film as photoanode for DSSC using natural fruit dyes.
Mohamad IS, et al.
AIP Conference Proceedings, 1660(1) (2015)
E Basoalto et al.
Environmental entomology, 39(5), 1399-1408 (2010-10-01)
We analyzed the spatial distribution and dispersal of codling moth, Cydia pomonella (L.), adults within two heterogeneous agroecosystems typical of central Chile: commercial apple, Malus domestica Borkhausen, orchards surrounded by various unmanaged host plants. Both a geostatistical analysis of catches
Monitoring of Chemical Speciation of DEA-CO2-Water System by Raman Spectroscopy.
Shahid MZ, et al.
Advanced Materials Research, 1113, 358-363 (2015)
Diethanolamine and N,N-diethylethanolamine ionic liquids as precatalyst-precursors and reaction media in green Heck reaction protocol.
Petrovic ZD, et al.
J. Mol. Catal. A: Chem., 327(1), 45-50 (2010)
Kelly K Ball et al.
Journal of neuroscience research, 89(12), 2052-2067 (2011-05-14)
Experimental diabetes increases production of reactive oxygen-nitrogen species and inhibits astrocytic gap junctional communication in tissue culture and brain slices from streptozotocin (STZ)-diabetic rats by unidentified mechanisms. Relative connexin (Cx) protein levels were assessed by Western blotting using extracts from

Protocols

Enzymatic Assay of Alkaline Phosphatase, Diethanolamine Assay (EC 3. 1. 3. 1)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service