Skip to Content
Merck
All Photos(1)

Documents

45323

Supelco

Amitraz

PESTANAL®, analytical standard

Synonym(s):

N′-(2,4-Dimethylphenyl)-N-{[(2,4-dimethylphenyl)imino]methyl}-N-methylmethanimidamide, NSC 324552

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H23N3
CAS Number:
Molecular Weight:
293.41
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

−20°C

SMILES string

CN(\C=N\c1ccc(C)cc1C)\C=N\c2ccc(C)cc2C

InChI

1S/C19H23N3/c1-14-6-8-18(16(3)10-14)20-12-22(5)13-21-19-9-7-15(2)11-17(19)4/h6-13H,1-5H3/b20-12+,21-13+

InChI key

QXAITBQSYVNQDR-ZIOPAAQOSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Prevention of tick borne diseases].
Marianne Bouman
Tijdschrift voor diergeneeskunde, 136(12), 898-898 (2012-01-13)
A Fernández-Salas et al.
Veterinary parasitology, 183(3-4), 338-342 (2011-08-10)
We document the presence of a Rhipicephalus microplus tick population resistant to acaricides (organophosphates (OP), synthetic pyrethroids (SP), amitraz) and macrocyclic lactones (ML) (ivermectin). Engorged females of R. microplus were collected from a cattle farm in Veracruz, Mexico, to evaluate
Frans Jongejan et al.
Veterinary parasitology, 179(4), 343-350 (2011-07-23)
Four groups of seven dogs were treated topically with a novel combination of fipronil, amitraz and (S)-methoprene in a spot-on formulation (CERTIFECT™, Merial Limited, GA, USA) on 28, 21, 14 and 7 days prior to tick infestation, respectively and acaricidal
Ekaterina Kuznetsova et al.
Veterinary parasitology, 188(1-2), 148-155 (2012-05-12)
Canine generalized demodicosis (CGD) is a skin disease with distinct breed predispositions. Secondary bacterial infections are common. Dogs typically receive miticidal therapy in combination with antibacterial treatment. Whether antibiotics influence the duration of acaricidal therapy is unknown at the moment.
Ralf S Mueller
Compendium (Yardley, PA), 34(4), E1-E4 (2012-04-11)
Canine demodicosis, a disease caused by a proliferation of Demodex mites, typically leads to alopecia, comedones, follicular papules and pustules, scaling, and crusting. It may be treated with either amitraz rinses or macrocyclic lactones. Amitraz rinse is approved for application

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service