Skip to Content
Merck
All Photos(1)

Documents

37924

Supelco

Thiamethoxam

PESTANAL®, analytical standard

Synonym(s):

3-(2-Chloro-5-thiazolylmethyl)tetrahydro-5-methyl-N-nitro-4H-1,3,5-oxadiazin-4-imine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10ClN5O3S
CAS Number:
Molecular Weight:
291.71
Beilstein:
8491021
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CN1COCN(Cc2cnc(Cl)s2)\C1=N\[N+]([O-])=O

InChI

1S/C8H10ClN5O3S/c1-12-4-17-5-13(8(12)11-14(15)16)3-6-2-10-7(9)18-6/h2H,3-5H2,1H3/b11-8+

InChI key

NWWZPOKUUAIXIW-DHZHZOJOSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Find more information here: Neonicotinoids
Thiamethoxam is a neonicotinoid broad spectrumand systemic insecticide, which belongs to the subclass of thianicotinyls. It can find applications in controlling the growth of sucking insects like aphids, whiteflies, and rice hoppers by acting on the acetylcholine nicotinic receptors.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Sol. 1 - Repr. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of nicotinic acetylcholine receptors from the insects Aphis craccivora, Myzus persicae, and Locusta migratoria by radioligand binding assays: relation to thiamethoxam action
Wiesner P and Kayser H
Journal of Biochemical and Molecular Toxicology, 14, 221-230 (2000)
P R Whitehorn et al.
Scientific reports, 7(1), 15562-15562 (2017-11-16)
Neonicotinoid pesticides have been linked to global declines of beneficial insects such as bumblebees. Exposure to trace levels of these chemicals causes sub-lethal effects, such as reduced learning and foraging efficiency. Complex behaviours may be particularly vulnerable to the neurotoxic
P Maienfisch et al.
Pest management science, 57(10), 906-913 (2001-11-07)
Thiamethoxam is the first commercial neonicotinoid insecticide from the thianicotinyl subclass. It was discovered in the course of our optimisation program on neonicotinoids started in 1985. Novel variations of the nitroimino-heterocycle of imidacloprid led to 4-nitroimino-1,3,5-oxadiazinanes exhibiting high insecticidal activity.
Effects of sublethal doses of acetamiprid and thiamethoxam on the behavior of the honeybee (Apis mellifera)
Hassani EKA, et al.
Archives of Environmental Contamination and Toxicology, 54, 653-661 (2008)
C Lobson et al.
The Science of the total environment, 637-638, 1150-1157 (2018-05-29)
Thiamethoxam is a neonicotinoid insecticide that can reach wetlands in agro-ecosystems through runoff. The fate and effects of thiamethoxam on non-target organisms in shallow wetland ecosystems have not been well characterized. To this end, a mesocosm study was conducted with

Protocols

Learn more about Neonicotinoids - active substances used in plant protection products to control harmful insects.

Learn more about Neonicotinoids - active substances used in plant protection products to control harmful insects.

Learn more about Neonicotinoids - active substances used in plant protection products to control harmful insects.

Learn more about Neonicotinoids - active substances used in plant protection products to control harmful insects.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service