Skip to Content
Merck
All Photos(1)

Documents

34176

Supelco

Oxfendazole

VETRANAL®, analytical standard

Synonym(s):

Methyl 5-(phenylsulfinyl)-benzimidazol-2-carbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H13N3O3S
CAS Number:
Molecular Weight:
315.35
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

COC(=O)Nc1nc2cc(ccc2[nH]1)S(=O)c3ccccc3

InChI

1S/C15H13N3O3S/c1-21-15(19)18-14-16-12-8-7-11(9-13(12)17-14)22(20)10-5-3-2-4-6-10/h2-9H,1H3,(H2,16,17,18,19)

InChI key

BEZZFPOZAYTVHN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Oxfendazole is a benzimidazole anthelmintic drug, effective against cysticercosis, echinococcosis and fasciolosis.
Oxfendazole is a member of benzimidazole group of anthelmintics.

Application

Oxfendazole may be used as a reference standard in the determination of oxfendazole in pharmaceutical preparations using reversed-phase high-performance liquid chromatography (RP-HPLC).
Oxfendazole may be used as an analytical reference standard for the determination of the analyte in bovine liver by liquid chromatography method.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1B - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C S Sikasunge et al.
Veterinary parasitology, 158(1-2), 57-66 (2008-10-07)
The aim of this study was to assess the effect of treating Taenia solium infected pigs with oxfendazole (OFZ) on viability and clearance of cysticerci and the corresponding persistence of specific antibody isotypes (IgG(total), IgG1, IgG2 and IgA) and circulating
L Moreno et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 50(10), 3819-3825 (2012-07-31)
Oxfendazole (OFZ) is efficacious for porcine cysticercosis at 30 mg/kg. OFZ is not registered to be used at this dose. The assessment of the OFZ and metabolites [(fenbendazole sulphone (FBZSO2), fenbendazole (FBZ)] plasma pharmacokinetic and tissue residue profiles after its
Yasuaki Dewa et al.
Archives of toxicology, 81(9), 647-654 (2007-03-07)
The effect of oxfendazole (OX), a benzimidazole anthelmintic, on hepatic gene expression was investigated in the liver of rats as a preliminary study to elucidate the possible mechanism of its non-genotoxic hepatocarcinogenesis. The liver from a male F344/N rat given
Jihei Nishimura et al.
Archives of toxicology, 84(2), 143-153 (2009-12-25)
To clarify whether enzymatically modified isoquercitrin (EMIQ) or melatonin (MLT) supplementation reduces oxidative stress-mediated hepatocellular tumor-promoting effect of oxfendazole (OX), a benzimidazole anthelmintic, male rats were administered a single intraperitoneal injection of N-diethylnitrosamine (DEN) and were fed a diet containing
Multi-residue method for the determination of benzimidazoles in bovine liver.
Dowling G, et al.
Analytica Chimica Acta, 529(1-2), 285-292 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service