Skip to Content
Merck
All Photos(2)

Key Documents

33818

Supelco

5-Hydroxythiabendazole

PESTANAL®, analytical standard

Synonym(s):

2-(4-Thiazolyl)-5-benzimidazolol, 5-Hydroxy-2-(4-thiazolyl)benzimidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H7N3OS
CAS Number:
Molecular Weight:
217.25
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental
forensics and toxicology
veterinary

format

neat

SMILES string

Oc1ccc2[nH]c(nc2c1)-c3cscn3

InChI

1S/C10H7N3OS/c14-6-1-2-7-8(3-6)13-10(12-7)9-4-15-5-11-9/h1-5,14H,(H,12,13)

InChI key

VNENJHUOPQAPAT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

5-Hydroxythiabendazole present in excreta, eggs, edible tissues and milk is found to be the major metabolite of thiabendazole, an active ingredient in fungicidal preparations effective for plants. It can also be used as an anthelmintic drug, which is applicable both for humans and animals.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Thiabendazole and its metabolite 5-hydroxythiabendazole can undergo specific binding with monoclonal antibodies and can further undergo immobilization on agarose gels, which can find applications in immunoaffinity chromatography.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G E Hardee et al.
Research in veterinary science, 43(1), 13-17 (1987-07-01)
Separate high performance liquid chromatographic methods were developed for thiabendazole (TBZ) and 5-hydroxy thiabendazole (5-OH-TBZ) determination in horse plasma using 1-methyl-2-phenyl benzimidazole (MPBZ) as an internal standard. In both methods TBZ and 5-OH-TBZ were extracted from plasma using organic solvents
Deepak Dalvie et al.
Drug metabolism and disposition: the biological fate of chemicals, 34(4), 709-717 (2006-01-26)
Thiabendazole (TBZ) is a broad-spectrum antihelmintic used for treatment of parasitic infections in animals and humans and as an agricultural fungicide for postharvest treatment of fruits and vegetables. It is teratogenic and nephrotoxic in mice, and cases of hepatotoxicity have
M Coulet et al.
Chemico-biological interactions, 127(2), 109-124 (2000-08-11)
Thiabendazole (TBZ), an anthelmintic and fungicide benzimidazole, was recently demonstrated to be extensively metabolized by cytochrome P450 (CYP) 1A2 in man and rabbit, yielding 5-hydroxythiabendazole (5OH-TBZ), the major metabolite furtherly conjugated, and two minor unidentified metabolites (M1 and M2). In
D Kerboeuf et al.
Veterinary parasitology, 93(1), 47-55 (2000-10-12)
Four groups of three lambs per group were experimentally infected with Cooperia curticei susceptible (two groups) or resistant (two groups) to benzimidazoles, and distributions of adult worms in the small intestine were studied. For each Cooperia isolate, one group was
Joseph D Jamieson et al.
Toxicology in vitro : an international journal published in association with BIBRA, 25(5), 1061-1066 (2011-04-26)
Thiabendazole (TBZ), an antihelminthic and antifungal agent, is associated with a host of adverse effects including nephrotoxicity, hepatotoxicity, and teratogenicity. Bioactivation of the primary metabolite of TBZ, 5-hydroxythiabendazole, has been proposed to yield a reactive intermediate. Here we show that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service