Skip to Content
Merck
All Photos(2)

Key Documents

35395

Supelco

Albendazole sulfoxide

VETRANAL®, analytical standard

Synonym(s):

Ricobendazole, Albendazole oxide, Albendazole sulfoxide, Methyl [5-(propane-1-sulfinyl)-1H-benzoimidazol-2-yl]-carbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15N3O3S
CAS Number:
Molecular Weight:
281.33
Beilstein:
677664
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

CCCS(=O)c1ccc2[nH]c(NC(=O)OC)nc2c1

InChI

1S/C12H15N3O3S/c1-3-6-19(17)8-4-5-9-10(7-8)14-11(13-9)15-12(16)18-2/h4-5,7H,3,6H2,1-2H3,(H2,13,14,15,16)

InChI key

VXTGHWHFYNYFFV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Ricobendazole is a key metabolite of albendazole and acts as an anthelmintic. It has been shown to induce apoptosis in human cancer cell line HT-29, possibly by arresting cell cycle at the G2/M phase.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 2 - Skin Sens. 1 - STOT RE 2 Oral

Target Organs

Adrenal gland,spleen,male reproductive organs,Blood

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Hai-Jun Gao et al.
Parasites & vectors, 14(1), 108-108 (2021-02-17)
Echinococcosis, which is caused by the larvae of cestodes of the genus Echinococcus, is a parasitic zoonosis that poses a serious threat to the health of humans and animals globally. Albendazole is the drug of choice for the treatment of
Viviane Cangerana Hilário et al.
Journal of pharmaceutical and biomedical analysis, 61, 100-107 (2012-01-11)
A high-performance liquid chromatographic method using polar organic mode was developed to analyze albendazole (ABZ), albendazole sulfone (ABZSO(2)) and the chiral and active metabolite albendazole sulfoxide (ABZSOX, ricobendazole) that was further applied in stereoselective fungal biotransformation studies. The chromatographic separation
Iliana González-Hernández et al.
Biomedical chromatography : BMC, 26(2), 267-272 (2011-07-02)
The development and validation of an LC-MS/MS method for the simultaneous determination of albendazole metabolites (albendazole sulfoxide and albendazole sulfone) in human plasma are described. Samples of 200 μL were extracted with ether-dichloromethane-chloroform (60:30:10, v/v/v). The chromatographic separation was performed
Tomasz Grabowski et al.
Biomedical chromatography : BMC, 25(10), 1159-1167 (2011-02-05)
Methods for determination of albendazole (ALB), albendazole sulfoxide (SOX) and albendazole sulfone (SON) in turkey blood plasma, using high-performance liquid chromatography (HPLC) with fluorescence detection, were developed. Moreover, comparison of HPLC columns with ultra-performance liquid chromatography (UPLC) columns was performed.
Kátia Roberta A Belaz et al.
Journal of pharmaceutical and biomedical analysis, 66, 100-108 (2012-04-11)
Analytical and semipreparative high performance liquid chromatography methods using polysaccharide-based chiral stationary phases were developed for the enantiomeric resolution of albendazol sulfoxide. The enantioseparation of this compound was evaluated with four chiral stationary phases: cellulose and amylose tris(3,5-dimethylphenylcarbamate), amylose tris[(S)-1-phenylethylcarbamate]

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service