Skip to Content
Merck
All Photos(2)

Key Documents

32499

Supelco

Dinotefuran

PESTANAL®, analytical standard

Synonym(s):

(RS)-N-Methyl-N′-nitro-N″-[(tetrahydro-3-furanyl)methyl]guanidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14N4O3
CAS Number:
Molecular Weight:
202.21
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CN\C(N[N+]([O-])=O)=N/CC1CCOC1

InChI

1S/C7H14N4O3/c1-8-7(10-11(12)13)9-4-6-2-3-14-5-6/h6H,2-5H2,1H3,(H2,8,9,10)

InChI key

YKBZOVFACRVRJN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dinotefuran, a new neonicotinoid insecticide has low mammalian toxicity and great insecticidal activity against a broad range of pests.
Find more information here: Neonicotinoids

Application

Dinotefuran has been used as a standard for the determination of neonicotinoid insecticide residues in dietary bee pollen and melon samples by HPLC–MS/MS and high performance liquid chromatography coupled with ultraviolet detector (HPLC-UVD), respectively.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raymond A Cloyd et al.
Pest management science, 67(1), 3-9 (2010-08-20)
The neonicotinoid insecticides imidacloprid, acetamiprid, dinotefuran, thiamethoxam and clothianidin are commonly used in greenhouses and/or interiorscapes (plant interiorscapes and conservatories) to manage a wide range of plant-feeding insects such as aphids, mealybugs and whiteflies. However, these systemic insecticides may also
Dinotefuran: a potential neonicotinoid insecticide against resistant mosquitoes.
Corbel V, et al.
Journal of Medical Entomology, 41(4), 712-717 (2004)
Frank J Byrne et al.
Pest management science, 68(5), 811-817 (2012-03-08)
Trunk injections of systemic insecticides were evaluated for the management of avocado thrips. Insecticide residues were quantified in leaves to determine when after treatment, and for how long, toxic concentrations of the insecticides were present. Residues in fruit were quantified
High-throughput HPLC?MS/MS determination of the persistence of neonicotinoid insecticide residues of regulatory interest in dietary bee pollen.
Lopez-Fernandez O, et al.
Analytical and Bioanalytical Chemistry, 407(23) (2015)
The discovery of dinotefuran: a novel neonicotinoid.
Wakita T, et al.
Pest Management Science, 59(9), 1016-1022 (2003)

Protocols

Learn more about Neonicotinoids - active substances used in plant protection products to control harmful insects.

Learn more about Neonicotinoids - active substances used in plant protection products to control harmful insects.

Learn more about Neonicotinoids - active substances used in plant protection products to control harmful insects.

Learn more about Neonicotinoids - active substances used in plant protection products to control harmful insects.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service