Skip to Content
Merck
All Photos(2)

Key Documents

37043

Supelco

Tolfenpyrad

PESTANAL®, analytical standard

Synonym(s):

4-Chloro-3-ethyl-1-methyl-N-[4-(p-tolyloxy)benzyl]pyrazole-5-carboxamide, 4-Chloro-3-ethyl-1-methyl-N-{[4-(4-methylphenoxy)phenyl]methyl}-1H-pyrazole-5-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H22ClN3O2
CAS Number:
Molecular Weight:
383.87
Beilstein:
13666608
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

SMILES string

CCc1nn(C)c(C(=O)NCc2ccc(Oc3ccc(C)cc3)cc2)c1Cl

InChI

1S/C21H22ClN3O2/c1-4-18-19(22)20(25(3)24-18)21(26)23-13-15-7-11-17(12-8-15)27-16-9-5-14(2)6-10-16/h5-12H,4,13H2,1-3H3,(H,23,26)

InChI key

WPALTCMYPARVNV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tolfenpyrad is an effective insecticide, used against pests that are resistant to existing insecticides such as organophosphates and carbamates.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of sample preparation on the estimation of residue levels of sprayed pesticides in separate analyses of turnip roots and leaves: inclusion or exclusion of the root-shoot junction
Yajima T, et al.
Journal of pest science, 2017 null
Tao Tang et al.
Journal of economic entomology, 113(6), 2793-2799 (2020-09-30)
The Asian citrus psyllid, Diaphorina citri Kuwayama, is the most serious pest of citrus because it is a vector for the highly destructive citrus greening disease (huanglongbing, HLB). Currently, insecticide applications are being used widely to control psyllid populations, thereby
Tao Tang et al.
Pest management science, 77(1), 168-176 (2020-07-12)
The Asian citrus psyllid (ACP), Diaphorina citri Kuwayama, is one of the most devastating pests in citrus orchards, and has caused huge economic losses worldwide. Chemical control is the most effective way for psyllid control. Herein, the toxicity of nine
Analysis of tolfenpyrad and its metabolites in plasma in a tolfenpyrad poisoning case
Yamaguchi K, et al.
Journal of Analytical Toxicology, 36, 529-537 (2012)
E R Echegaray et al.
Journal of economic entomology, 110(1), 142-149 (2016-12-29)
Bactericera cockerelli (Šulc) is a common pest of solanaceous crops largely known for vectoring "psyllid yellows" in potatoes. In recent years, however, this pest has attracted considerable attention for vectoring Zebra Chip, a devastating bacterial disease that was first reported

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service