Skip to Content
Merck
All Photos(2)

Documents

215643

Sigma-Aldrich

Disperse Blue 1

Dye content 30 %

Synonym(s):

1,4,5,8-Tetraaminoanthraquinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H12N4O2
CAS Number:
Molecular Weight:
268.27
Colour Index Number:
64500
Beilstein:
925672
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

powder

composition

Dye content, 30%

mp

332 °C

solubility

1 M NH4OH: 10 mg/mL, opaque, blue

λmax

607 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Nc1ccc(N)c2C(=O)c3c(N)ccc(N)c3C(=O)c12

InChI

1S/C14H12N4O2/c15-5-1-2-6(16)10-9(5)13(19)11-7(17)3-4-8(18)12(11)14(10)20/h1-4H,15-18H2

InChI key

JSFUMBWFPQSADC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Disperse Blue 1 is a grey to black powder. Disperse dyes are known to be insoluble in water and are applied to the substrate as suspensions or dispersions by dissolving in hydrophobic regions of the polymers making up the fiber.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

A Cruz et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 44(4), 159-166 (2001-09-29)
A sequential anaerobic/aerobic treatment process was used to biodegrade the azo dye disperse blue 79 (DB79). The integrated process was successfully tested and operated for the mineralization of the DB79 dye until a concentration of 120 mg/l in 96 h.
Disperse Blue 1.
IARC monographs on the evaluation of carcinogenic risks to humans, 48, 139-148 (1990-01-01)
Nat. Toxicology Program
Report on Carcinogens (12th Ed. ) (2011)
L C Haws et al.
Regulatory toxicology and pharmacology : RTP, 19(1), 80-96 (1994-02-01)
Disperse Blue 1 is an anthraquinone dye used at low levels in semipermanent hair color formulations. Dietary administration of Disperse Blue 1 in a National Toxicology Program (NTP) carcinogenesis bioassay produced transitional- and squamous-cell tumors, leiomyomas, and leiomyosarcomas of the
Yang Liu et al.
Journal of medicinal chemistry, 52(21), 6523-6526 (2009-10-21)
A series of mitoxantrone (MTX) analogues have been designed, synthesized, and evaluated for binding to and stabilizing a stem-loop structure that serves as a splicing regulatory element in the pre-mRNA of tau, which is involved in Alzheimer's and other neurodegenerative

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service