Skip to Content
Merck
All Photos(2)

Documents

A39009

Sigma-Aldrich

1-Aminoanthraquinone

≥96.5% purity, powder, crystals or chunks

Synonym(s):

Azoic Diazo No. 36, Fast Red AL

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H9NO2
CAS Number:
Molecular Weight:
223.23
Colour Index Number:
37275
Beilstein:
396360
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

product name

1-Aminoanthraquinone, ≥96.5% (HPLC)

Quality Level

Assay

≥96.5% (HPLC)

form

powder, crystals or chunks

color

dark brown

mp

253-255 °C (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Nc1cccc2C(=O)c3ccccc3C(=O)c12

InChI

1S/C14H9NO2/c15-11-7-3-6-10-12(11)14(17)9-5-2-1-4-8(9)13(10)16/h1-7H,15H2

InChI key

KHUFHLFHOQVFGB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

1-Aminoanthraquinone has the ability to increase the solubility of derivatives of anthraquinone in supercritical carbon dioxide.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Solubility of 1-aminoanthraquinone and 1-nitroanthraquinone in supercritical carbon dioxide
Tamura K, et al.
The Journal of Chemical Thermodynamics, 104, 162-168 (2017)
Song Zhang et al.
Scientific reports, 7, 43419-43419 (2017-02-25)
We investigated the mechanism of intramolecular charge transfer and the following radiationless dynamics of the excited states of 1-aminoanthraquinone using steady state and time-resolved absorption spectroscopy combined with quantum chemical calculations. Following photoexcitation with 460 nm, conformational relaxation via twisting of
Jin-Shui Liu et al.
Guang pu xue yu guang pu fen xi = Guang pu, 25(1), 76-78 (2005-04-28)
Anthracene, perylene and 1-aminoanthraquinone colloids have been prepared by reprecipitation method. In comparison with the fluorescence emission spectra of colloids and molecules (in acetone solution), it was indicated that collids fluorescence spectra are red-shifted and easy emission. Base on the
A Klimek-Turek et al.
Journal of chromatography. A, 1530, 204-210 (2017-11-25)
A concept of using thin-layer chromatography to multicomponent sample preparation for quantitative determination of solutes followed by instrumental technique is presented. Thin-layer chromatography (TLC) is used to separate chosen substances and their internal standard from other components (matrix) and to
S Pflugmacher et al.
The Science of the total environment, 357(1-3), 169-175 (2005-05-12)
Dissolved natural organic matter (NOM) is dead organic matter exceeding, in freshwater systems, the concentration of organic carbon in all living organisms by far. 80-90% (w/w) of the NOM is made up of humic substances (HS). Although NOM possesses several

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service