Skip to Content
Merck
All Photos(2)

Documents

367842

Sigma-Aldrich

1,5-Diaminoanthraquinone

technical grade, 85%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10N2O2
CAS Number:
Molecular Weight:
238.24
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

Assay

85%

form

powder

mp

>300 °C (lit.)

SMILES string

Nc1cccc2C(=O)c3c(N)cccc3C(=O)c12

InChI

1S/C14H10N2O2/c15-9-5-1-3-7-11(9)14(18)8-4-2-6-10(16)12(8)13(7)17/h1-6H,15-16H2

InChI key

VWBVCOPVKXNMMZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,5-Diaminoanthraquinone (DAAQ) is an organic dye compound. Rapid preparation of 1,5-diaminoanthraquinone nanofibers (DAAQNFs) decorated with small platinum nanoparticles (PtNPs) has been reported. The absorption and fluorescence spectra of DAAQ has been investigated in various organic solvents. Preparation of vertical organic nanowire arrays of DAAQ on solid substrates by a facile physical vapor transport method has been reported.

Application

1,5-Diaminoanthraquinone (DAAQ) may be used in the preparation of vertical nanowire arrays. It may be used in the synthesis of poly(1,5-diaminoanthraquinone) (PDAA) particles, via chemically oxidative polymerization of DAAQ.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Suganthi et al.
Journal of fluorescence, 20(1), 95-103 (2009-08-27)
The absorption and fluorescence spectra of 1,5-diaminoanthraquinone(1,5-DAAQ) have been investigated in organic solvents-Benzene(BZ), Ethanol (ETOH), Acetonitrile (AN), Dimethylformamide (DMF) and Dimethyl sulfoxide (DMSO). There is an intra molecular hydrogen bond formed between quinoid oxygen and the substituents NH(2) [C =
Xin-Gui Li et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 13(31), 8884-8896 (2007-07-27)
Wholly aromatic poly(1,5-diaminoanthraquinone) (PDAA) particles were productively synthesized for the first time by chemically oxidative polymerization of 1,5-diaminoanthraquinone (DAA) by using CrO3, K2Cr2O7, K2CrO4, or KMnO4 as oxidants in acidic DMF. The effects of the oxidant species, oxidant/DAA ratio, polymerization
Yong Sheng Zhao et al.
ACS nano, 4(3), 1630-1636 (2010-02-11)
Vertical nanowire arrays were prepared from an organic dye compound, 1,5-diaminoanthraquinone (DAAQ), on various types of substrates by a facile physical vapor transport method. It was found that the DAAQ grows much faster on the substrates with higher surface energies.
Yong Sheng Zhao et al.
Journal of the American Chemical Society, 131(9), 3158-3159 (2009-03-05)
Vertical organic nanowire arrays of 1,5-diaminoanthraquinone (DAAQ) on solid substrates were prepared by a facile physical vapor transport method. The crystal structure and growth direction of the nanowires were determined with electron diffraction. The color and fluorescence of the DAAQ
Guohui Chang et al.
Journal of nanoscience and nanotechnology, 12(9), 7075-7080 (2012-10-06)
The present communication reports on the rapid preparation of 1,5-diaminoanthraquinone nanofibers (DAAQNFs) on a large scale by a reprecipitation method and their subsequent decoration with small platinum nanoparticles (PtNPs) using tannic acid (TA) as a reducing agent. It suggests the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service