Skip to Content
Merck
All Photos(1)

Key Documents

73285

Supelco

Butyl acetate

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COO(CH2)3CH3
CAS Number:
Molecular Weight:
116.16
Beilstein:
1741921
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

4 (vs air)

vapor pressure

15 mmHg ( 25 °C)
8 mmHg ( 20 °C)

Assay

≥99.7% (GC)

autoignition temp.

790 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

7.6 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.394 (lit.)
n20/D 1.394

bp

124-126 °C (lit.)

mp

−78 °C (lit.)

density

0.88 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCCCOC(C)=O

InChI

1S/C6H12O2/c1-3-4-5-8-6(2)7/h3-5H2,1-2H3

InChI key

DKPFZGUDAPQIHT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Butyl acetate can find wide applications in painting and coating industries. It can also serve as a substitute to toxic solvents used in the manufacture of acrylic polymers, enamels, vinyl resins, nitrocellulose and lacquers. It is generally prepared by the esterification of butanol with acetic acid in the presence of sulfuric acid. It can also be obtained via transesterification reaction of methyl acetate with butanol.
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Find all available reference materials for compounds listed in 10/2011 here

Application

Butyl acetate may be used as an analytical standard for the determination of the analyte in paint solvents by spectrophotometry, and in amorolfine liniment and carbamate pesticides by chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

80.6 °F - closed cup

Flash Point(C)

27 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

A hybrid reactive distillation process with high selectivity pervaporation for butyl acetate production via transesterification
Harvianto RG, et al.
Journal of Membrane Science, 543, 49-57 (2017)
Application of liquid-phase microextraction and on-column derivatization combined with gas chromatography-mass spectrometry to the determination of carbamate pesticides
Zhang J and Lee HK
Journal of Chromatography A, 1117(1), 31-37 (2006)
Simultaneous vapour phase Fourier transform infrared spectrometric determination of butyl acetate, toluene and methyl ethyl ketone in paint solvents
Lopez-Anreus E
Analyst, 123(6), 1247-1252 (1998)
Simultaneous determination of triacetin, acetic ether, butyl acetate and amorolfine hydrochloride in amorolfine liniment by HPLC.
Gao Y, et al.
Pakistan Journal of Pharmaceutical Sciences, 25(2) (2012)
Hala Fam et al.
Bioprocess and biosystems engineering, 35(8), 1367-1374 (2012-03-29)
The mass transfer of phenol and butyl acetate to/from water was studied in two-phase partitioning bioreactors using immiscible organic solvents and solid polymer beads as the partitioning phases in a 5-L stirred tank bioreactor. Virtually instantaneous mass transfer was observed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service