Skip to Content
Merck
All Photos(1)

Key Documents

18616

Sigma-Aldrich

Kaolin

heavy, powder

Synonym(s):

Aluminum silicate hydroxide, Bolus, Hydrated aluminum silicate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
~Al2Si2O5(OH)4
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
11111803
NACRES:
NA.25
form:
powder

form

powder

Quality Level

quality

heavy

impurities

acid. or alk. react. impurities, in accordance
organic impurities, in accordance
≤0.01% heavy metals (as Pb)
≤1% soluble in acid (as SO4)

capacity

in accordance absorption capacity

anion traces

chloride (Cl-): ≤250 mg/kg
sulfate (SO42-): ≤1000 mg/kg

cation traces

Ca: ≤250 mg/kg

suitability

in accordance for identity
passes test for swelling capacity

application(s)

pharmaceutical (small molecule)

SMILES string

[Si](=O)(O[Si](=O)O[Al]=O)O[Al]=O.O.O

InChI

1S/2Al.O5Si2.2H2O.2O/c;;1-6(2)5-7(3)4;;;;/h;;;2*1H2;;/q2*+1;-2;;;;

InChI key

NLYAJNPCOHFWQQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ching-Wei Wu et al.
Journal of hazardous materials, 244-245, 412-420 (2013-01-01)
The process of mechanical milling has been found to effectively stabilize heavy metals in municipal solid waste incinerator (MSWI) fly ash, as well as to restrain the evaporation of heavy metals during thermo-treatment. This method is adopted in this study
Kenji Inaba et al.
The journal of trauma and acute care surgery, 74(2), 538-545 (2013-01-29)
The purpose of this study was to evaluate the long-term efficacy and safety of kaolin- and chitosan-based hemostatic agents for hemorrhage control in a 14-day survival, damage-control swine model of Grade IV liver injury. A total of 48 anesthetized pigs
Nicholas R Kunio et al.
American journal of surgery, 205(5), 505-510 (2013-03-19)
The purpose of this study was to compare standard gauze (SG) and advanced hemostatic dressings in use by military personnel in a no-hold model. A randomized, controlled trial was conducted using 36 swine. Animals underwent femoral arteriotomy, followed by 60
Salvador Cotillas et al.
Water research, 47(5), 1741-1750 (2013-01-29)
In this work, a novel integrated electrochemical process for urban wastewater regeneration is described. The electrochemical cell consists in a Boron Doped Diamond (BDD) or a Dimensionally Stable Anode (DSA) as anode, a Stainless Steel (SS) as cathode and a
B C A Pinheiro et al.
Journal of environmental management, 118, 205-210 (2013-03-05)
This study investigates the incorporation of solid petroleum waste as raw material into a porcelain stoneware tile body, in replacement to natural kaolin material by up to 5 wt.%. Tile formulations containing solid petroleum waste were pressed and fired at 1240 °C

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service