Merck
  • Mechanochemically induced synthesis of anorthite in MSWI fly ash with kaolin.
Mechanochemically induced synthesis of anorthite in MSWI fly ash with kaolin.
Journal of hazardous materials (2013-01-01)
Ching-Wei Wu, Chang-Jung Sun, Sue-Huai Gau, Cheng-Lin Hong, Cheng-Gang Chen
ABSTRACT
The process of mechanical milling has been found to effectively stabilize heavy metals in municipal solid waste incinerator (MSWI) fly ash, as well as to restrain the evaporation of heavy metals during thermo-treatment. This method is adopted in this study and the composition and degree of amorphization adjusted to improve the efficiency of crystalline anorthite synthesis. Different milling times (1, 5, 10 and 20 h) and different sintering temperatures (900, 950, 1000, 1100, 1200 and 1300 °C) are utilized. The extracted fly ash and kaolin (KEFA) were mixed to simulate an anorthite composite. The experimental results indicate that the degree of amorphization of the KEFA increased as the milling time increased. Furthermore, the synthesis of crystalline anorthite increased as the degree of amorphization increased. The milling process allowed a reduction in the synthesization temperature from 1300 °C to 950 °C. The heavy metals are sealed in during the liquid sintering phase, which reduces the amount of heavy metals released from the sintered specimens.
Sigma-Aldrich
Kaolin, heavy, powder
Sigma-Aldrich
Halloysite nanoclay
Sigma-Aldrich
Kaolin, meets USP testing specifications
Sigma-Aldrich
Kaolin, tested according to Ph. Eur.
Sigma-Aldrich
Kaolin