Skip to Content
Merck
All Photos(1)

Key Documents

69904

Sigma-Aldrich

Montmorillonite

K 30

Synonym(s):

K-Catalyst

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.22

Quality Level

form

powder

surface area

~330 m2/g

pH

2.8-3.8

SMILES string

[Si](=O)=O.[Si](=O)=O.[Si](=O)=O.[Si](=O)=O.[Al+3].[Al+3].[O-2].[O-2].[O-2].O

InChI

1S/2Al.4O2Si.H2O.3O/c;;4*1-3-2;;;;/h;;;;;;1H2;;;/q2*+3;;;;;;3*-2

InChI key

GUJOJGAPFQRJSV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

catalyst for alkylation reactions, rearrangements and isomerizations

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 3

1 of 3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service