Skip to Content
Merck
All Photos(4)

Key Documents

185450

Sigma-Aldrich

Phenol

unstabilized, ReagentPlus®, ≥99%

Synonym(s):

Hydroxybenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5OH
CAS Number:
Molecular Weight:
94.11
Beilstein:
969616
EC Number:
MDL number:
UNSPSC Code:
12352104
eCl@ss:
39023301
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

vapor density

3.24 (vs air)

Quality Level

vapor pressure

0.09 psi ( 55 °C)
0.36 mmHg ( 20 °C)

product line

ReagentPlus®

Assay

≥99%

form

crystalline (flakes)
crystalline (solid)
crystals

autoignition temp.

1319 °F

expl. lim.

8.6 %

bp

182 °C (lit.)

mp

40-42 °C (lit.)

solubility

water: soluble 84 g/L at 20 °C

density

1.071 g/mL at 25 °C (lit.)

SMILES string

Oc1ccccc1

InChI

1S/C6H6O/c7-6-4-2-1-3-5-6/h1-5,7H

InChI key

ISWSIDIOOBJBQZ-UHFFFAOYSA-N

Gene Information

human ... GABRA1(2554)

Looking for similar products? Visit Product Comparison Guide

General description

Phenol is an aromatic alcohol. Various industrial processes for the synthesis of phenol have been proposed.[1] It can undergo oxidation in the presence of Fentons′ reagent in aqueous solution.[2]

Application

Phenol may be employed in the following studies:
  • Isolation of deoxyribonucleic acid from Bacillus subtilis.[3]
  • Berthelot color reaction for the estimation of ammonia.[4]
  • Preparation of phenetole (aromatic ether)[5] and 1,2-dihydroxybenzene (catechol).[6]
  • Synthesis of phenoxyacetic acid.[7]
Phenol may be used in the following processes:
  • As a phase-transfer catalyst in electrolyte solutions of lithium-O2 battery to promote solution phase discharge.[8]
  • Preparation of phenol-sulfuric acid reagent for colorimetric determination of total sugar content.[9]

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - STOT RE 2

Target Organs

Nervous system,Kidney,Liver,Skin

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

177.8 °F - closed cup

Flash Point(C)

81 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Carbohydrate analysis by a phenol-sulfuric acid method in microplate format.
Masuko T, et al.
Analytical Biochemistry, 339(1), 69-72 (2005)
Chemical pathway and kinetics of phenol oxidation by Fenton's reagent.
Zazo JA, et al.
Environmental Science & Technology, 39(23), 9295-9302 (2005)
Phenol-Catalyzed Discharge in the Aprotic Lithium-Oxygen Battery.
Gao X, et al.
Angewandte Chemie (Weinheim an der Bergstrasse, Germany) (2017)
Eagleson M.
Concise Encyclopedia Chemistry, 812-812 (1994)
Tyman JHP.
Synthetic and Natural Phenols., 67-67 (1996)

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service