Skip to Content
Merck
All Photos(1)

Key Documents

P5566

Sigma-Aldrich

Phenol

BioXtra, ≥99.5% (GC)

Synonym(s):

Hydroxybenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5OH
CAS Number:
Molecular Weight:
94.11
Beilstein:
969616
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.31

vapor density

3.24 (vs air)

Quality Level

vapor pressure

0.09 psi ( 55 °C)
0.36 mmHg ( 20 °C)

product line

BioXtra

Assay

≥99.5% (GC)

form

powder

autoignition temp.

1319 °F

contains

0.15% H3PO2 as stabilizer

expl. lim.

8.6 %

ash

≤0.05%

color

white

bp

182 °C (lit.)

mp

40-42 °C (lit.)

solubility

H2O: 0.5 M at 20 °C, clear, colorless

density

1.071 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.0005%
Ba: ≤0.0005%
Bi: ≤0.0005%
Ca: ≤0.001%
Cd: ≤0.0005%
Co: ≤0.0005%
Cr: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Li: ≤0.0005%
Mg: ≤0.0005%
Mn: ≤0.0005%
Mo: ≤0.0005%
Na: ≤0.005%
Ni: ≤0.0005%
Pb: ≤0.0005%
Sr: ≤0.0005%
Zn: ≤0.0005%

storage temp.

2-8°C

SMILES string

Oc1ccccc1

InChI

1S/C6H6O/c7-6-4-2-1-3-5-6/h1-5,7H

InChI key

ISWSIDIOOBJBQZ-UHFFFAOYSA-N

Gene Information

human ... GABRA1(2554)

Looking for similar products? Visit Product Comparison Guide

General description

Phenol is a predominant pollutant in the environment. Phenol is also used in cosmetic and pharmaceutical industry. Phenol pollutes water bodies and inhibits the γ-amino butyric acid receptor (GABAAR)-coupled chloride ATPase function in the fish brain.

Application

Phenol has been used:
  • in the preparation chromotrope 2R staining solution.
  • as a reagent in α-amylase inhibitory activity.
  • as a standard in calibration of NMR spectra.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - STOT RE 2

Target Organs

Nervous system,Kidney,Liver,Skin

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of phenol on the GABAAR-coupled Cl-/HCO3-ATPase from fish brain: An in vitro approach on the enzyme function
Menzikov SA
Toxicology in vitro, 46, 129-136 (2018)
Recent advances in the development of biosensor for phenol: a review
Karim F and Fakhruddin ANM
Reviews in Environmental Science and BioTechnology, 11(3), 261-274 (2012)
Development of a Method for Determining the Amino Acid Composition of the Pharmaceutical Substance ?Glatiramer Acetate? by NMR Spectroscopy
Kuzmina NE, et al.
Pharmaceutical Chemistry Journal, 51(3), 226-229 (2017)
Comparison of four staining methods for detecting eosinophils in nasal polyps
Song Y, et al.
Scientific Reports, 8(1), 17718-17718 (2018)
Bioactive Phenolics of the Genus Artemisia (Asteraceae): HPLC-DAD-ESI-TQ-MS/MS Profile of the Siberian Species and Their Inhibitory Potential Against alpha-Amylase and alpha-Glucosidase
Olennikov DN, et al.
Frontiers in Pharmacology, 9 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service