Skip to Content
Merck
All Photos(3)

Key Documents

179981

Sigma-Aldrich

Luperox® A98, Benzoyl peroxide

reagent grade, ≥98%

Synonym(s):

Benzoyl peroxide, Dibenzoyl peroxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5CO)2O2
CAS Number:
Molecular Weight:
242.23
Beilstein:
984320
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

grade

reagent grade

Quality Level

Assay

≥98%

form

solid

autoignition temp.

176 °F

reaction suitability

reagent type: oxidant

mp

105 °C (lit.)

functional group

peroxide
phenyl

SMILES string

O=C(OOC(=O)c1ccccc1)c2ccccc2

InChI

1S/C14H10O4/c15-13(11-7-3-1-4-8-11)17-18-14(16)12-9-5-2-6-10-12/h1-10H

InChI key

OMPJBNCRMGITSC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzoyl peroxide participates in the covalent insitu functionalization of carbon nanotubes. On heating, it undergoes decomposition to afford phenyl free radicals and CO2 gas.
Luperox® A98, Benzoyl peroxide, an anhydrous benzoyl peroxide powder, is a mainly used as a thermal free radical initiator in polymerization reactions.

Application

Benzoyl peroxide has been used as an initiator in the preparation of the following:
  • Thermal cross-linking of multiwalled carbon nanotubes (MWCNTs) via radical-initiated reaction
  • 3-D crosslinked carbon scaffolds
  • poly(lactic-co-glycolic) acid (PLGA), MWCNT and singlewalled carbon nanotubes (SWCNT) scaffolds
Widely used initiator, curing agent, and cross-linking agent in polymerization processes.

Legal Information

Product of Arkema Inc.
Luperox is a registered trademark of Arkema Inc.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Org. Perox. B - Skin Sens. 1A

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gaurav Lalwani et al.
Carbon, 53, 90-100 (2013-02-26)
We report a simple method to fabricate macroscopic, 3-D, free standing, all-carbon scaffolds (porous structures) using multiwalled carbon nanotubes (MWCNTs) as the starting materials. The scaffolds prepared by radical initiated thermal crosslinking, and annealing of MWCNTs possess macroscale interconnected pores
E J Kang et al.
Journal of food science, 77(7), C818-C823 (2012-07-05)
Residual annatto colorant (norbixin) in fluid Cheddar cheese whey can be bleached. The 2 approved chemical bleaching agents for whey, hydrogen peroxide (HP) and benzoyl peroxide (BP), negatively impact the flavor of dried whey protein. The objective of this study
Suzanne M Jervis et al.
Journal of food science, 78(2), R129-R137 (2013-01-22)
Whey is a value-added product that is utilized in many food and beverage applications for its nutritional and functional properties. Whey and whey products are generally utilized in dried ingredient applications. One of the primary sources of whey is from
Adam J Friedman et al.
The Journal of investigative dermatology, 133(5), 1231-1239 (2012-11-30)
Advances in nanotechnology have demonstrated potential application of nanoparticles (NPs) for effective and targeted drug delivery. Here we investigated the antimicrobial and immunological properties and the feasibility of using NPs to deliver antimicrobial agents to treat a cutaneous pathogen. NPs
A L Kraus et al.
Regulatory toxicology and pharmacology : RTP, 21(1), 87-107 (1995-02-01)
Topical benzoyl peroxide has been used in the treatment of acne for over 30 years, with no reports of adverse effects that could be related to skin carcinogenesis. Two case-control epidemiological studies have found a lack of association between the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service