Skip to Content
Merck
All Photos(4)

Key Documents

290785

Sigma-Aldrich

Luperox® LP, Lauroyl peroxide

≥98%

Synonym(s):

Lauroyl peroxide, Di(dodecanoyl) peroxide, DiDodecanoyl Peroxide, Dilauroyl peroxide, Dodecanoyl peroxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)10CO]2O2
CAS Number:
Molecular Weight:
398.62
Beilstein:
1804936
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor density

13.7 (vs air)

Assay

≥98%

form

solid

reaction suitability

reagent type: oxidant

mp

53-57 °C (lit.)

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCC(=O)OOC(=O)CCCCCCCCCCC

InChI

1S/C24H46O4/c1-3-5-7-9-11-13-15-17-19-21-23(25)27-28-24(26)22-20-18-16-14-12-10-8-6-4-2/h3-22H2,1-2H3

InChI key

YIVJZNGAASQVEM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Lauroyl peroxide can be used as:
  • A long chain branching(LCB) introducer in polypropylene to improve its mechanical recycling process.
  • A mild oxidant and efficient hydrogen abstractor in the preparation of disulfide compounds through dimerization of corresponding dithiocarbamates.
  • An initiator in the preparation of ion-imprinted polymers(IIPs).
  • An initiator and oxidant to construct erythrina ring system via oxidative radical cyclizations of enamide in the presence of nBu3SnH.

Legal Information

Product of Arkema Inc.
Luperox is a registered trademark of Arkema Inc.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Org. Perox. D

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Saeed Kakaei et al.
Organic & biomolecular chemistry, 11(33), 5481-5490 (2013-07-17)
A series of (2-alkylthiothiazolin-5-yl)methyl dodecanoates was synthesized from various alkyl N-allylcarbamodithioates and dilauroyl peroxide via a tandem radical hydrogen-abstraction-cyclization-substitution/combination reaction with a 5-exo-trig radical cyclization as a key step. The current route is the first, convenient, and efficient synthesis of
A J Klein-Szanto et al.
The Journal of investigative dermatology, 79(1), 30-34 (1982-07-01)
Free radical generating peroxides are potent skin irritants. After a single topical application of either 10, 20, or 40 mg of lauroyl peroxides or benzoyl peroxide on the dorsal skin of Sencar mice, the epidermal thickness increased markedly. No major
Y Sorata et al.
Biochimica et biophysica acta, 799(3), 313-317 (1984-06-29)
Photosensitized hemolysis of human erythrocytes by hematoporphyrin was suppressed by flavonols such as quercetin and rutin at submillimolar concentrations. The suppression of photohemolysis was accompanied by inhibition of lipid peroxidation by the reagents. Quercetin and rutin were photooxidized in the
Studies on the effect of functional monomer and porogen on the properties of ion imprinted polymers based on Cr(III)-1,10-phenanthroline complex designed for selective removal of Cr(III) ions
Laura Trzonkowska, et al.
Reactive and Functional Polymers, 117, 131-139 (2017)
Itxaso Parola et al.
Polymers, 11(7) (2019-07-18)
This work reports on a diameter dependence analysis of the performance as luminescent solar concentrators of three self-fabricated polymer optical fibers (POFs) doped with a hybrid combination of dopants. The works carried out include the design and self-fabrication of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service