Skip to Content
Merck
All Photos(1)

Key Documents

B5907

Sigma-Aldrich

Benzoyl peroxide blend with dicyclohexyl phthalate

suitable for use as a catalyst for electron microscopy. Modified to render it safe in transit.

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10O4
Molecular Weight:
242.23
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Quality Level

form

powder

contains

50% dicyclohexyl phthalate as stabilizer

reaction suitability

reagent type: oxidant

packaging

vial of 9.9 g

mp

54 °C

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

SMILES string

O=C(OOC(=O)c1ccccc1)c2ccccc2

InChI

1S/C14H10O4/c15-13(11-7-3-1-4-8-11)17-18-14(16)12-9-5-2-6-10-12/h1-10H

InChI key

OMPJBNCRMGITSC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Linkage

Formulated specifically for use with Histocryl and LR white acrylic resins.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Org. Perox. D - Repr. 1B - Skin Sens. 1

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Laszlo Prokai et al.
Science translational medicine, 7(297), 297ra113-297ra113 (2015-07-24)
Many neurological and psychiatric maladies originate from the deprivation of the human brain from estrogens. However, current hormone therapies cannot be used safely to treat these conditions commonly associated with menopause because of detrimental side effects in the periphery. The
Meital Bloch et al.
International journal of pharmaceutics, 478(2), 504-516 (2014-12-02)
To increase colonoscopy capability to discriminate benign from malignant polyps, we suggest combining two imaging approaches based on targeted polymeric platforms. Water-soluble cationized polyacrylamide (CPAA) was tagged with the near infrared (NIR) dye IR-783-S-Ph-COOH to form Flu-CPAA. The recognition peptide
Megan E Oest et al.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society, 33(3), 334-342 (2014-11-20)
Late-onset fragility fractures are a common complication following radiotherapy for metastatic disease and soft tissue sarcomas. Using a murine hindlimb focal irradiation model (RTx), we quantified time-dependent changes in osteoclasts and mineral apposition rate (MAR). Mice received either a single
Ahmed M Debela et al.
Biosensors & bioelectronics, 71, 25-29 (2015-04-17)
An alternative strategy for surface tethering of DNA probes, where highly reactive glassy carbon (GC) substrates are prepared via electrochemical hydrogenation and electrochemical/chemical chlorination is reported. Thiolated DNA probes and alkanethiols were stably immobilised on the halogenated carbon, with electrochemical
Hemanth Akkiraju et al.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society, 33(2), 208-215 (2014-10-22)
Bone Morphogenetic Protein 2 (BMP2) regulates bone integrity by driving both osteogenesis and osteoclastogenesis. However, BMP2 as a therapeutic has significant drawbacks. We have designed a novel peptide CK2.3 that blocks the interaction of Casein Kinase 2 (CK2) with Bone

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service