Skip to Content
Merck
All Photos(2)

Key Documents

14211

Supelco

Water

ACS reagent, CHROMASOLV®, suitable for HPLC and ultratrace analysis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2O
CAS Number:
Molecular Weight:
18.02
Beilstein:
2050024
EC Number:
MDL number:
UNSPSC Code:
12191602
PubChem Substance ID:
NACRES:
NA.24
grade:
ACS reagent
for inorganic trace analysis
technique(s):
immunofluorescence: suitable
application(s):
sample preparation
bp:
100 °C (lit.)

Product Name

Water, for ultratrace analysis, ACS reagent

grade

ACS reagent
for inorganic trace analysis

Quality Level

form

liquid

technique(s)

immunofluorescence: suitable

impurities

≤5 μg/kg ammonium (NH4)

refractive index

n20/D 1.34 (lit.)

pH

6.0-8.0(-25 °C77 °F)

bp

100 °C (lit.)

mp

0 °C (lit.)

solubility

water: miscible (completely)

density

1.000 g/mL at 3.98 °C (lit.)

anion traces

bromide (Br-): ≤1 μg/kg
chloride (Cl-): ≤1 μg/kg
fluoride (F-): ≤1 μg/kg
iodide (I-): ≤1 μg/kg
nitrate (NO3-): ≤1 μg/kg
nitrite (NO2-): ≤1 μg/kg
phosphate (PO43-): ≤1 μg/kg
sulfate (SO42-): ≤1 μg/kg

cation traces

Ag: ≤0.001 μg/kg
Al: ≤0.5 μg/kg
As: ≤0.01 μg/kg
Au: ≤0.005 μg/kg
B: ≤0.5 μg/kg
Ba: ≤0.5 μg/kg
Be: ≤0.005 μg/kg
Bi: ≤0.001 μg/kg
Ca: ≤0.5 μg/kg
Cd: ≤0.5 μg/kg
Ce: ≤0.001 μg/kg
Co: ≤0.5 μg/kg
Cr: ≤0.5 μg/kg
Cs: ≤0.001 μg/kg
Cu: ≤0.5 μg/kg
Dy: ≤0.001 μg/kg
Er: ≤0.001 μg/kg
Eu: ≤0.001 μg/kg
Fe: ≤0.5 μg/kg
Ga: ≤0.001 μg/kg
Gd: ≤0.001 μg/kg
Ge: ≤0.005 μg/kg
Hf: ≤0.001 μg/kg
Hg: ≤0.5 μg/kg
Ho: ≤0.001 μg/kg
In: ≤0.001 μg/kg
Ir: ≤0.001 μg/kg
K: ≤0.5 μg/kg
La: ≤0.001 μg/kg
Li: ≤0.5 μg/kg
Lu: ≤0.001 μg/kg
Mg: ≤0.5 μg/kg
Mn: ≤0.5 μg/kg
Mo: ≤0.5 μg/kg
Na: ≤0.5 μg/kg
Nb: ≤0.005 μg/kg
Nd: ≤0.001 μg/kg
Ni: ≤0.005 μg/kg
Os: ≤0.005 μg/kg
Pb: ≤05 μg/kg
Pd: ≤0.001 μg/kg
Pr: ≤0.001 μg/kg
Pt: ≤0.005 μg/kg
Rb: ≤0.001 μg/kg
Re: ≤0.005 μg/kg
Rh: ≤0.001 μg/kg
Ru: ≤0.005 μg/kg
Sb: ≤0.001 μg/kg
Sc: ≤0.005 μg/kg
Se: ≤5 μg/kg
Si: ≤0.5 μg/kg
Sm: ≤0.001 μg/kg
Sn: ≤0.5 μg/kg
Sr: ≤0.5 μg/kg
Ta: ≤0.001 μg/kg
Tb: ≤0.001 μg/kg
Te: ≤0.001 μg/kg
Th: ≤0.005 μg/kg
Ti: ≤0.5 μg/kg
Tl: ≤0.005 μg/kg
Tm: ≤0.001 μg/kg
U: ≤0.001 μg/kg
V: ≤0.5 μg/kg
W: ≤0.005 μg/kg
Y: ≤0.001 μg/kg
Yb: ≤0.001 μg/kg
Zn: ≤0.5 μg/kg
Zr: ≤0.5 μg/kg

SMILES string

O

application(s)

sample preparation

InChI

1S/H2O/h1H2

InChI key

XLYOFNOQVPJJNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Susu Duan et al.
Nature communications, 5, 5029-5029 (2014-10-10)
Oseltamivir-resistant H1N1 influenza viruses carrying the H275Y neuraminidase mutation predominated worldwide during the 2007-2009 seasons. Although several neuraminidase substitutions were found to be necessary to counteract the adverse effects of H275Y, the order and impact of evolutionary events involved remain
Mary A Rodgers et al.
The Journal of experimental medicine, 211(7), 1333-1347 (2014-06-25)
Linear ubiquitination is a newly discovered posttranslational modification that is currently restricted to a small number of known protein substrates. The linear ubiquitination assembly complex (LUBAC), consisting of HOIL-1L, HOIP, and Sharpin, has been reported to activate NF-κB-mediated transcription in
Boris Nowka et al.
Applied and environmental microbiology, 81(2), 745-753 (2014-11-16)
Nitrification has an immense impact on nitrogen cycling in natural ecosystems and in wastewater treatment plants. Mathematical models function as tools to capture the complexity of these biological systems, but kinetic parameters especially of nitrite-oxidizing bacteria (NOB) are lacking because
Yasushi Itoh et al.
PLoS pathogens, 10(6), e1004192-e1004192 (2014-06-20)
Highly pathogenic avian influenza (HPAI) viruses of the H5N1 subtype often cause severe pneumonia and multiple organ failure in humans, with reported case fatality rates of more than 60%. To develop a clinical antibody therapy, we generated a human-mouse chimeric
M Grusdat et al.
Cell death and differentiation, 21(7), 1050-1060 (2014-02-18)
CD8(+) T-cell functions are critical for preventing chronic viral infections by eliminating infected cells. For healthy immune responses, beneficial destruction of infected cells must be balanced against immunopathology resulting from collateral damage to tissues. These processes are regulated by factors

Articles

Learn more about Bioethanol as a renewable, alternative fuel traditionally produced by the yeast fermentation of sugar.

Learn more about Bioethanol as a renewable, alternative fuel traditionally produced by the yeast fermentation of sugar.

Learn more about Bioethanol as a renewable, alternative fuel traditionally produced by the yeast fermentation of sugar.

Learn more about Bioethanol as a renewable, alternative fuel traditionally produced by the yeast fermentation of sugar.

Protocols

Separation of Methane; Acetylene; Carbon monoxide; Water; Nitrogen; Carbon dioxide; Ethane; Ethylene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service