Skip to Content
Merck
All Photos(1)

Key Documents

12309

Supelco

p-Benzoquinone

for spectrophotometric det. of amines, ≥99.5% (HPLC)

Synonym(s):

Quinone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(=O)2
CAS Number:
Molecular Weight:
108.09
Beilstein:
773967
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

vapor density

3.73 (vs air)

Quality Level

vapor pressure

0.1 mmHg ( 25 °C)

Assay

≥99.5% (HPLC)

form

powder or crystals

autoignition temp.

815 °F

quality

for spectrophotometric det. of amines

technique(s)

UV/Vis spectroscopy: suitable

sublimation residue

≤0.1%

color

, Faint Yellow to Very Dark Yellow to Very Dark Beige and Green-Yellow

mp

112-114 °C
113-115 °C (lit.)

SMILES string

O=C1C=CC(=O)C=C1

InChI

1S/C6H4O2/c7-5-1-2-6(8)4-3-5/h1-4H

InChI key

AZQWKYJCGOJGHM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

P-Benzoquinone along with tetrathiafulvalene (TTF) was used as mediators, in an experimental study based on different parameters affecting the preparation of enzyme electrochemical biosensors using self-assembled monolayer (SAM)-modified electrodes is reported.[1] It may be used for spectrophotometric determination of cysteine and/or carbocysteine in a mixture of amino acids, shampoo, and pharmaceutical.[2] It may also be used as an effective cogent in peroxide-initiated crosslinking of polypropylene.[3]

Other Notes

Reagent for the spectrophotometric microdetermination of amines[4]

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Flam. Sol. 1 - Muta. 2 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

170.6 °F - closed cup

Flash Point(C)

77 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peroxide-initiated crosslinking of polypropylene in the presence of p-benzoquinone.
Chodak I and Lazar M.
Journal of Polymer Science: Part A, General Papers, 32 (6), 5432-5437 (1986)
Spectrophotometric microdetermination of amines and sulfamates with 1,4-Benzoquinone.
G A Benson et al.
Analytical chemistry, 48(14), 2149-2152 (1976-12-01)
D A Zaia et al.
Talanta, 50(5), 1003-1010 (2008-10-31)
A simple, sensitive, and selective method has been developed for determination of cysteine (Cys) or carbocysteine (carboCys) in pharmaceutical products, shampoos and a mixture of amino acids. The results showed the reaction between p-benzoquinone (PBQ) and Cys occurs through the
Preparation, characterization and application of alkanethiol self-assembled monolayers modified with tetrathiafulvalene and glucose oxidase at a gold disk electrode.
Campuzano S
Journal of Electroanalytical Chemistry, 526 (1), 92-100 (2002)
Rozbeh Baradaran et al.
Nature, 494(7438), 443-448 (2013-02-19)
Complex I is the first and largest enzyme of the respiratory chain and has a central role in cellular energy production through the coupling of NADH:ubiquinone electron transfer to proton translocation. It is also implicated in many common human neurodegenerative

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service