Skip to Content
Merck

Skip To

8.02410

p-Benzoquinone

for synthesis

Synonym(s):

p-Benzoquinone, p-Quinone, 1,4-Benzoquinone, 1,4-Cyclohexadiene-3,6-dione

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C6H4O2
CAS Number:
Molecular Weight:
108.09
UNSPSC Code:
12352115
EC Index Number:
203-405-2
NACRES:
NA.22
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

0.12 hPa ( 20 °C)

Quality Segment

form

solid

autoignition temp.

560 °C

potency

130 mg/kg LD50, oral (Rat)

pH

4 (20 °C, 1 g/L in H2O)

mp

110-113 °C

solubility

10 g/L

density

1.32 g/cm3 at 20 °C

bulk density

700 kg/m3

storage temp.

2-30°C

SMILES string

O=C1C=CC(=O)C=C1

InChI

1S/C6H4O2/c7-5-1-2-6(8)4-3-5/h1-4H

InChI key

AZQWKYJCGOJGHM-UHFFFAOYSA-N

Application

p-Benzoquinone can be used as:
  • A free-radical inhibitor.
  • A catalyst to synthesize highly site-selective N1-alkylated benzotriazoles by N1-alkylation of benzotriazoles with diazo compounds.
  • A hydrogen acceptor and two electron oxidant in Pd-catalyzed Wacker oxidation of aryl olefins aldehydes.
  • A redox mediator in Pd-catalyzed anaerobic electrooxidative homocoupling of aryl-boron derivatives.


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
802940104610195464
description

for synthesis

description

for synthesis

description

Technipur®

description

98%

mp

110-113 °C

mp

209-214 °C

mp

172-175 °C (lit.)

mp

235 °C (dec.) (lit.)

vapor pressure

0.12 hPa ( 20 °C)

vapor pressure

-

vapor pressure

1 mmHg ( 132 °C)

vapor pressure

-

solubility

10 g/L

solubility

-

solubility

-

solubility

-

autoignition temp.

560 °C

autoignition temp.

-

autoignition temp.

930 °F

autoignition temp.

-

potency

130 mg/kg LD50, oral (Rat)

potency

86 mg/kg LD50, oral (Rat)

potency

-

potency

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

100


Still not finding the right product?

Explore all of our products under p-Benzoquinone


signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Flam. Sol. 1 - Muta. 2 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

4.1B - Flammable solid hazardous materials

wgk

WGK 3

flash_point_f

170.6 °F - closed cup

flash_point_c

77 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters