Skip to Content
Merck
All Photos(3)

Key Documents

114367

Sigma-Aldrich

Bromocresol Green sodium salt

ACS reagent, 90% (HPLC), Dye content 90 %

Synonym(s):

3′,3″,5′,5″-Tetrabromo-m-cresolsulfonphthalein sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H13Br4NaO5S
CAS Number:
Molecular Weight:
720.00
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

grade

ACS reagent

Quality Level

Agency

suitable for SM 2310
suitable for SM 2320

Assay

90% (HPLC)

form

powder or crystals

composition

Dye content, 90%

visual transition interval

3.8, yellow(Visual Interval Pass)
3.8-5.2, yellow to blue
4.5, green
5.4, blue(Visual Interval Pass)

mp

230 °C (dec.) (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].Cc1c(Br)c(O)c(Br)cc1\C(=C2\C=C(Br)C(=O)C(Br)=C2C)c3ccccc3S([O-])(=O)=O

InChI

1S/C21H14Br4O5S.Na/c1-9-12(7-14(22)19(26)17(9)24)21(13-8-15(23)20(27)18(25)10(13)2)11-5-3-4-6-16(11)31(28,29)30-21;/h3-8,26-27H,1-2H3;/q;+1/p-1

InChI key

HEFSGAHJDGZCHA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Bromocresol Green (BCG), also known as bromcresol green, is a synthetic, anionic, triphenylmethane dye belonging to the phthalein and sulphonphthalein dyes group. It is commonly used as a pH indicator as it changes color from yellow to blue-green at pH 3.8-5.4. The monosodium salt of bromocresol green is highly ionizable in water. The dye at a higher pH level produces a large and weakly lipophilic dianion.

Application

Bromocresol green serves as a pH indicator in biology and medicinal experiments.
  • It is used widely as a vital stain to study blood-brain permeability.
  • It is used as a tracking dye for DNA agarose gel electrophoresis, in protein determinations, and in charge-transfer complexation processes.
  • It is used in TLC for the visualization of compounds with functional groups whose pKa is below 5.0.
  • It has been used in the pH-sensitive colorimetric determination of glutamate decarboxylase activity.
  • It has also been employed in the evaluation of paper-analytical devices for colorimetric analysis of serum samples.
  • It is suitable as a selective inhibitor in kidney physiology studies.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Conn?s Biological Stains: A Handbook of Dyes, Stains and Fluorochromes for Use in Biology and Medicine (10th ed.)
RW Horobin, JA Kiernan (Eds.)
Conn?s Biological Stains (2002)
A high-throughput colorimetric assay to measure the activity of glutamate decarboxylase
Yu, et al.
Enzyme and Microbial Technology, 49(3) (2011)
Azo-functionalized superparamagnetic Fe3O4 nanoparticles: an efficient adsorbent for the removal of bromocresol green from contaminated water
Saad, et al.
Royal Society of Chemistry Advances, 12 (2022)
Selection of appropriate protein assay method for a paper microfluidics platform
Pokhrel, at al.
practical laboratory medicine, 21 (2020)
Martina O' Toole et al.
Talanta, 66(5), 1340-1344 (2008-10-31)
A novel inexpensive optical-sensing technique has been developed for colorimetric flow analysis. This sensing system employs two LEDs whereby one is used as the light source and the other as a light detector. The LED used as light detector is

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service