Skip to Content
Merck
All Photos(3)

Key Documents

861367

Sigma-Aldrich

Thymol Blue sodium salt

ACS reagent, Dye content 95 %

Synonym(s):

Thymolsulfonphthalein sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H29NaO5S
CAS Number:
Molecular Weight:
488.57
Beilstein:
3861515
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

grade

ACS reagent

Quality Level

form

powder or crystals

composition

Dye content, 95%

clarity of soln

passes test

visual transition interval

1.2-2.8, red to yellow(acid range)
8.0-9.2, yellow to blue(alkaline range)

mp

283-285 °C (dec.) (lit.)

λmax

378 nm (2nd)
590 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].CC(C)c1cc(c(C)cc1O)C2(OS(=O)(=O)c3ccccc23)c4cc(C(C)C)c([O-])cc4C

InChI

1S/C27H30O5S.Na/c1-15(2)19-13-22(17(5)11-24(19)28)27(21-9-7-8-10-26(21)33(30,31)32-27)23-14-20(16(3)4)25(29)12-18(23)6;/h7-16,28-29H,1-6H3;/q;+1/p-1

InChI key

BAVBEHWEOJMHDS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Douglas R Macfarlane et al.
Chemical communications (Cambridge, England), 46(41), 7703-7705 (2010-09-28)
We describe hydrated ionic liquids which have an intrinsic proton buffering action and hence an ability to control proton activity in an ionic medium.
N Takai et al.
Artificial organs, 15(2), 86-89 (1991-04-01)
An optical fiber sensor utilizing Thymol blue and an ion-exchange resin complex in a cellulose acetate membrane was developed. By monitoring several different chromophores of Thymol blue, the sensor could measure the pH of the solution from 1.0 to 12.0
Shangjie Xu et al.
Macromolecular bioscience, 7(8), 968-974 (2007-08-01)
A simple general synthetic concept to build dendritic core-shell architectures with pH-labile linkers based on hyperbranched PEI cores and biocompatible PEG shells is presented. Using these dendritic core-shell architectures as nanocarriers, the encapsulation and transport of polar dyes of different
N Gayathri et al.
Journal of AOAC International, 84(4), 1065-1069 (2001-08-15)
A simple and sensitive spectrophotometric method was developed for the determination of trace amounts of sulfur dioxide. The method is based on the reaction of SO2 with a known excess of ICI as the oxidant. The unreacted ICI iodinates thymol
P Balderas-Hernández et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 66(1), 68-73 (2006-07-11)
This paper presents the results concerning the determination of the formation constants of the complexes between thymol blue, TB, and the metal ions Pb(II) or Cd(II). The experimental procedure was carried out in the presence of a nitrogen atmosphere at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service