Skip to Content
Merck
All Photos(2)

Key Documents

B1256

Sigma-Aldrich

Bromocresol Green sodium salt

≥98% purity, crystalline

Synonym(s):

3′,3″,5′,5″-Tetrabromo-m-cresolsulfonphthalein sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H13Br4NaO5S
CAS Number:
Molecular Weight:
720.00
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

product name

Bromocresol Green sodium salt, crystalline

Agency

suitable for SM 2310
suitable for SM 2320

Quality Level

form

crystalline

visual transition interval

3.8-5.4, yellow to blue

mp

230 °C (dec.) (lit.)

solubility

H2O: 1 mg/mL

λmax

616 nm in H2O

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].Cc1c(Br)c(O)c(Br)cc1\C(=C2\C=C(Br)C(=O)C(Br)=C2C)c3ccccc3S([O-])(=O)=O

InChI

1S/C21H14Br4O5S.Na/c1-9-12(7-14(22)19(26)17(9)24)21(13-8-15(23)20(27)18(25)10(13)2)11-5-3-4-6-16(11)31(28,29)30-21;/h3-8,26-27H,1-2H3;/q;+1/p-1

InChI key

HEFSGAHJDGZCHA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Bromocresol Green sodium salt has been used as a pH indicator for glutamate decarboxylase activity assay in Listeria monocytogenes strains.

Biochem/physiol Actions

Bromocresol Green is mainly used for measuring serum albumin. In presence of acidic conditions, bromocresol green interacts with albumin, thereby resulting in an increase in absorbance at 630nm. The change in absorbance is linked with the concentration of albumin. Bromocresol Green is also a pH indicator, changing from yellow (pH 4.0) to blue color (5.4).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Screening of glutamate decarboxylase activity and bile salt resistance of human asymptomatic carriage, clinical, food, and environmental isolates of Listeria monocytogenes.
International Journal of Food Microbiology, 93, 87-99 (2004)
Remington: The Science and Practice of Pharmacy (2006)
Homocysteine as a predictive biomarker in early diagnosis of renal failure susceptibility and prognostic diagnosis for end stages renal disease.
Amin HK, et al.
Renal Failure, 38, 1267-1275 (2016)

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service