Skip to Content
Merck
All Photos(1)

Key Documents

09415

Sigma-Aldrich

Sodium 4-aminosalicylate dihydrate

≥97.0% (T)

Synonym(s):

4-Amino-2-hydroxybenzoic acid sodium salt, 4-Amino-salicylic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6NNaO3 · 2H2O
CAS Number:
Molecular Weight:
211.15
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.0% (T)

form

powder

reaction suitability

reaction type: solution phase peptide synthesis

technique(s)

ligand binding assay: suitable

color

white to faintly beige

application(s)

peptide synthesis

SMILES string

O.O.[Na+].Nc1ccc(c(O)c1)C([O-])=O

InChI

1S/C7H7NO3.Na.2H2O/c8-4-1-2-5(7(10)11)6(9)3-4;;;/h1-3,9H,8H2,(H,10,11);;2*1H2/q;+1;;/p-1

InChI key

GMUQJDAYXZXBOT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S M Vesenbeckh et al.
Infection, 40(2), 199-202 (2011-08-13)
para-Aminosalicylic acid (PAS) is commonly used in the treatment of drug-resistant tuberculosis, including multidrug-resistant tuberculosis. Since its first use in the 1940s, hypersensitivity reactions frequently limit its use in clinical practice. Cases of successful desensitization against PAS using orally administered
Shuichi Yano et al.
BMJ case reports, 2012, doi:10-doi:10 (2012-05-19)
Isoniazid (H) or rifampicin (R) mono-resistant disease can be treated easily and effectively with first-line drugs, while combined H and R resistance (ie, multidrug-resistant tuberculosis (MDRTB)) requires treatment with at least four agents, including a quinolone and an injectable agent.
V E Sagynbaeva et al.
Eksperimental'naia i klinicheskaia gastroenterologiia = Experimental & clinical gastroenterology, (3)(3), 40-46 (2012-07-27)
Detection of IgM and IgG Chlamydia and Mycoplasma pneumoniae indicates an aggravation of intracellular infections, including, possibly, due to immunosuppressive therapy. It is possible that the intracellular infection may mediate the occurrence of certain extraintestinal manifestations of inflammatory bowel disease
[A chronic moderate manganism poisoning treated with sodium p-aminosalicylic acid].
Wei-Ping Qin et al.
Zhonghua lao dong wei sheng zhi ye bing za zhi = Zhonghua laodong weisheng zhiyebing zazhi = Chinese journal of industrial hygiene and occupational diseases, 29(8), 619-620 (2012-02-18)
Lan Hong et al.
Drug metabolism and disposition: the biological fate of chemicals, 39(10), 1904-1909 (2011-07-20)
para-aminosalicylic acid (PAS; 4-amino-2-hydroxybenzoic acid), an antituberculosis drug in use since the 1950s, has recently been suggested to be an effective agent for treatment of manganese-induced parkinsonian disorders. However, the neuropharmacokinetics of PAS and its metabolite N-acetyl-para-aminosalicylic acid (AcPAS; N-acetyl-4-amino-2-hydroxybenzoic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service