Skip to Content
Merck
All Photos(3)

Key Documents

E4630

Sigma-Aldrich

Ethambutol dihydrochloride

antimycobacterial

Synonym(s):

Ethambutol, Ethambutol HCL, 2,2′-(1,2-Ethanediyldiimino)bis-1-butanol dihydrochloride, Emb

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H24N2O2 · 2HCl
CAS Number:
Molecular Weight:
277.23
EC Number:
MDL number:
UNSPSC Code:
51281511
PubChem Substance ID:
NACRES:
NA.85

form

powder

optical activity

[α]25/D 6.0 to 6.7°, c = 10 in water

color

white

solubility

H2O: 50 mg/mL

antibiotic activity spectrum

mycobacteria

Mode of action

cell wall synthesis | interferes

SMILES string

CC[C@@H](CO)NCCN[C@H](CO)CC.Cl.Cl

InChI

1S/C10H24N2O2.2ClH/c1-3-9(7-13)11-5-6-12-10(4-2)8-14;;/h9-14H,3-8H2,1-2H3;2*1H/t9-,10-;;/m0../s1

InChI key

AUAHHJJRFHRVPV-BZDVOYDHSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ethambutol is an antimycobacterial used to treat tuberculosis in combination with other antibiotics to reduce the development of drug resistance.
Ethambutol is an antimycobacterial used to treat tuberculosis in combination with other antibiotics to reduce the development of drug resistance. It has been used to study multidrug-resistant tuberculosis (MDR-TB) in highest-incidence-rate areas in China.

Biochem/physiol Actions

Ethambutol is bacteriostatic against actively growing TB bacilli. Mycolic acids attach to the 5′-hydroxyl groups of D-arabinose residues of arabinogalactan and form mycolyl-arabinogalactan-peptidoglycan complex in the cell wall. Ethambutol disrupts arabinogalactan synthesis by inhibiting arabinosyltransferases, specifically those in Mycobacterium, interfering with the biosynthesis of arabinogalactans found in the cell wall of mycobacteria and leading to increased cell wall permeability.
Ethambutol specifically inhibits arabinosyltransferases in Mycobacterium, interfering with the biosynthesis of arabinogalactans found in the cell wall of mycobacteria.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hassan Safi et al.
Nature genetics, 45(10), 1190-1197 (2013-09-03)
To study the evolution of drug resistance, we genetically and biochemically characterized Mycobacterium tuberculosis strains selected in vitro for ethambutol resistance. Mutations in decaprenylphosphoryl-β-D-arabinose (DPA) biosynthetic and utilization pathway genes Rv3806c, Rv3792, embB and embC accumulated to produce a wide
Hao Zeng et al.
International journal of clinical and experimental medicine, 8(6), 8398-8414 (2015-08-27)
To introduce a modified method, namely coaxial electrohydrodynamic atomization for the fabrication of distinct core/shell structured microspheres of four first-line ant-tuberculosis drugs with different characteristics in hydrophilic properties in one single step. In group B, we prepared microspheres in which
A case of skin and lung tuberculosis: diagnostic difficulties.
P Colasanti et al.
Giornale italiano di dermatologia e venereologia : organo ufficiale, Societa italiana di dermatologia e sifilografia, 148(5), 538-540 (2013-09-06)
Joshua R Francis et al.
The Medical journal of Australia, 200(6), 328-332 (2014-04-08)
OBJECTIVE To describe the epidemiology, clinical features, health care resource use, treatment and outcomes of multidrug-resistant tuberculosis (MDR-TB) cases diagnosed in Western Australia, compared with matched controls with drug-susceptible TB. Retrospective case-control study of all MDR-TB cases notified in WA
N Guo et al.
Letters in applied microbiology, 58(4), 344-349 (2013-12-18)
Previous studies have shown that isoimperatorin (IO), a furanocoumarin isolated from several medicinal plants, has antimycobacterial activity against Mycobacterium tuberculosis strain H37Rv (ATCC 27294). This study demonstrated that IO has antimycobacterial activity against 2 drug-sensitive and 6 drug-resistant isolates, with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service