Skip to Content
Merck
All Photos(1)

Key Documents

D9754

Sigma-Aldrich

2-Aminoethyl diphenylborinate

97%

Synonym(s):

2-APB, 2-Aminoethoxydiphenyl borate, 2-Aminoethyl diphenyl borate, Diphenylboric acid 2-aminoethyl ester, Diphenylborinic acid, NSC 17107

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2BOCH2CH2NH2
CAS Number:
Molecular Weight:
225.09
Beilstein:
2942273
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

storage temp.

−20°C

SMILES string

NCCOB(c1ccccc1)c2ccccc2

InChI

1S/C14H16BNO/c16-11-12-17-15(13-7-3-1-4-8-13)14-9-5-2-6-10-14/h1-10H,11-12,16H2

InChI key

BLZVCIGGICSWIG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Preparation of apoptotic inducers

  • Activator of the Orai3 (ORAI calcium release-activated calcium modulator 3) protein
  • Inhibits Ca2+ influx through transient receptor potential 2 channels in rat dorsal root ganglion neurons
  • Membrane-permeable SOCE (store operated calcuym entry) inhibitor

Biochem/physiol Actions

Membrane-permeable modulator of intracellular IP3-induced calcium release.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Akinobu Z Suzuki et al.
Bioorganic & medicinal chemistry letters, 20(4), 1395-1398 (2010-01-26)
Store-operated calcium entry (SOCE) is an important mechanism for replenishing intracellular calcium stores and for sustaining calcium signaling. We developed a method for synthesis of bisboron compounds that have two borinic acids or their esters in one molecule. These compounds
Mustafa Nazıroğlu et al.
The Journal of membrane biology, 241(2), 69-75 (2011-04-22)
Exposure to oxidative stress causes health problems, including sensory neuron neuropathy and pain. Rotenone is a toxin used to generate intracellular oxidative stress in neurons. However, the mechanism of toxicity in dorsal root ganglion (DRG) neurons has not been characterized.
T Maruyama et al.
Journal of biochemistry, 122(3), 498-505 (1997-11-05)
The effects of a novel membrane-penetrable modulator, 2APB (2-aminoethoxy diphenyl borate), on Ins(1,4,5)P3-induced Ca2+ release were examined. 2APB inhibited Ins(1,4,5)P3-induced Ca2+ release from rat cerebellar microsomal preparations without affecting [3H]Ins(1,4,5)P3 binding to its receptor. The IC50 value (concentration producing 50%
Velasco-Bejarano, B.; et al.
Synlett, 921-921 (2007)
Judith Bergsmann et al.
The Journal of biological chemistry, 286(36), 31565-31575 (2011-07-05)
STIM1 and Orai represent the key components of Ca(2+) release-activated Ca(2+) channels. Activation of Orai channels requires coupling of the C terminus of STIM1 to the N and C termini of Orai. Although the latter appears to be central in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service