Skip to Content
Merck
All Photos(1)

Key Documents

Y0001568

Isoquercitroside

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H20O12
CAS Number:
Molecular Weight:
464.38
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

isoquercitroside

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O1[C@H]([C@@H]([C@H]([C@@H]([C@H]1CO)O)O)O)Oc2[c](c3c([o]c2c4cc(c(cc4)O)O)cc(cc3O)O)=O

InChI

1S/C21H20O12/c22-6-13-15(27)17(29)18(30)21(32-13)33-20-16(28)14-11(26)4-8(23)5-12(14)31-19(20)7-1-2-9(24)10(25)3-7/h1-5,13,15,17-18,21-27,29-30H,6H2/t13-,15-,17+,18-,21+/m1/s1

InChI key

OVSQVDMCBVZWGM-QSOFNFLRSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Isoquercitroside EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toshiaki Makino et al.
Journal of natural medicines, 67(4), 881-886 (2013-03-16)
Quercetin, a flavonol distributed widely in edible and medicinal plants, has beneficial biological activities in vitro. However, the low water solubility of quercetin limits its bioavailability to exhibit activity in vivo. We evaluated the anti-allergic effects of quercetin, quercetin 3-O-glucoside
Madeline J Simpson et al.
Molecules (Basel, Switzerland), 18(9), 11264-11280 (2013-09-17)
The anti-peroxyl radical quality of two aqueous rooibos infusions and solutions of their most abundant glycosylated polyphenols was evaluated using pyrogallol red and fluorescein-based oxygen radical absorbance ratios. It was observed that the artificial infusions, prepared using only the most
Linling Lu et al.
Biomedical chromatography : BMC, 27(4), 509-514 (2012-09-29)
In this paper, ultraperformance liquid chromatography/quadrupole time-of-flight mass spectrometry (UPLC-Q-TOF/MS) and the MetaboLynx™ software combined with mass defect filtering were applied to identity the metabolites of isoquercitrin using an intestinal mixture of bacteria and 96 isolated strains from human feces.
Lin Liu et al.
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials, 36(1), 132-136 (2013-06-12)
To Set up a method for determining the contents of the five flavonoids simultaneously in the HuangKui capsule and analyze their specific chromatograms. HPLC method was used. The analytical column was Thermo scientific Hypersil GOLD (250 mm x 4.6 mm
Jianyong Zhang et al.
Journal of ethnopharmacology, 148(2), 563-569 (2013-05-21)
Atherosclerosis (AS) can result in severe cardiovascular diseases. Early indications of AS include disorders in lipid metabolism, inflammatory responses, and endothelial dysfunction. Statins are the preferred drugs for stabilizing atherosclerotic plaques because of their lipid-lowering, anti-inflammation and endothelial-protection activities. However

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service