Skip to Content
Merck
All Photos(1)

Key Documents

24-0380

Sigma-Aldrich

Paraffin wax

SAJ first grade, mp 52-54 °C

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12000000

grade

SAJ first grade

form

solid

availability

available only in Japan

mp

52-54 °C

InChI key

JWHAUXFOSRPERK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

602.6 °F - Pensky-Martens closed cup

Flash Point(C)

317 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roman Zimmermann et al.
Plant physiology, 152(1), 356-365 (2009-11-21)
In an approach to study lateral root development in monocots, genome-wide searches for homologs of the Gibberellic Acid Stimulated Transcript-like (GAST-like) gene family in rice (Oryza sativa) and maize (Zea mays) were carried out. Six novel GAST-like genes in rice
Wisam Al-Faqheri et al.
PloS one, 8(3), e58523-e58523 (2013-03-19)
This paper introduces novel vacuum/compression valves (VCVs) utilizing paraffin wax. A VCV is implemented by sealing the venting channel/hole with wax plugs (for normally-closed valve), or to be sealed by wax (for normally-open valve), and is activated by localized heating
Yu Seob Shin et al.
Urology, 81(2), 437-441 (2013-02-05)
To introduce a new repair technique, inverted V-shape, for penile paraffinoma without necrosis of the ventral skin anastomosed. From March 2006 to July 2012, 34 patients underwent penile paraffinoma repair using the T-style or a new operation procedure, the inverted
Abdulazeez Salawu et al.
PloS one, 7(11), e50415-e50415 (2012-12-05)
Most soft tissue sarcomas are characterized by genetic instability and frequent genomic copy number aberrations that are not subtype-specific. Oligonucleotide microarray-based Comparative Genomic Hybridisation (array CGH) is an important technique used to map genome-wide copy number aberrations, but the traditional
JunYi Soh et al.
Journal of clinical pathology, 66(4), 312-318 (2013-02-09)
Fourier transform infrared (FT-IR) imaging is increasingly being applied to biomedical specimens, but strong IR absorption by water complicates live cell imaging. This study investigates the viability of adherent epithelial cells maintained for short periods under mineral oils in order

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service