Skip to Content
Merck
All Photos(1)

Key Documents

18512

Sigma-Aldrich

Paraffin oil

puriss., meets analytical specification of Ph. Eur., BP, viscous liquid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12191502
NACRES:
NA.21

Agency

meets analytical specifications of BP
meets analytical specifications of Ph. Eur.

Quality Level

grade

puriss.

form

viscous liquid

quality

meets analytical specification of Ph. Eur., BP

technique(s)

IR spectroscopy: suitable

impurities

acidity or alkalinity, complies
aromatic polycycl. hydrocarbons, complies
residual solvents, complies
solid parafins, complies

dynamic viscosity

110-230 mPa.s

density

0.827-0.890 g/mL at 20 °C

suitability

complies for reaction against H2SO4
in accordance for identity (IR)

application(s)

metabolomics
pharmaceutical (small molecule)
vitamins, nutraceuticals, and natural products

Looking for similar products? Visit Product Comparison Guide

Application


  • Polymer Physics behind the Gel-Spinning of UHMWPE Fibers: Though indirectly related, the study′s focus on material science can be paralleled with research on paraffin oil′s role in polymer processing and synthesis, especially in contexts where its lubricating properties are critical to the manufacturing processes (Wang et al., 2024).

Other Notes

The article number 18512-4X2.5L will be discontinued. Please order the single bottle 18512-2.5L which is physically identical with the same exact specifications.
The article number 18512-6X1L will be discontinued. Please order the single bottle 18512-1L which is physically identical with the same exact specifications.
Pertains only in Germany: unversteuert

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

419.0 °F - closed cup

Flash Point(C)

215 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Stefan N Sanders et al.
Biomechanics and modeling in mechanobiology, 19(5), 1585-1594 (2020-01-26)
Accurate estimation of mechanical properties of the different atherosclerotic plaque constituents is important in assessing plaque rupture risk. The aim of this study was to develop an experimental set-up to assess material properties of vascular tissue, while applying physiological loading
Nicole Concin et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 46(2), 544-552 (2007-10-10)
Paraffins of mineral oil origin (mineral paraffins) were analyzed in tissue fat collected from 144 volunteers with Caesarean sections as well as in milk fat from days 4 and 20 after birth of the same women living in Austria. In
Wenming Wu et al.
The Analyst, 136(11), 2287-2293 (2011-04-05)
Bubble formation inside a microscale channel is a significant problem in general microfluidic experiments. The problem becomes especially crucial when performing a polymerase chain reaction (PCR) on a chip which is subject to repetitive temperature changes. In this paper, we
Yi-Shiung Horng et al.
American journal of physical medicine & rehabilitation, 90(6), 435-442 (2011-03-25)
: The aim of this study was to investigate the effectiveness of tendon and nerve gliding exercises as a part of combined treatments for carpal tunnel syndrome. : Patients with carpal tunnel syndrome were randomized into three groups. All patients
Jyothi C Abbar et al.
Bioelectrochemistry (Amsterdam, Netherlands), 83, 1-7 (2011-07-26)
The electrochemical oxidation of a hemorheologic drug, pentoxifylline was investigated at a multi-walled carbon nanotubes-paraffin oil paste electrode using cyclic and differential pulse voltammetry. The oxidation process was irreversible over the pH range studied and exhibited an adsorption-controlled behavior. All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service