Skip to Content
Merck
All Photos(1)

Key Documents

07-3750

Sigma-Aldrich

Diiodomethane

SAJ first grade, ≥98.0%

Synonym(s):

Methylene iodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH2I2
CAS Number:
Molecular Weight:
267.84
Beilstein:
1696892
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

grade

SAJ first grade

vapor density

9.25 (vs air)

Assay

≥98.0%

form

liquid

availability

available only in Japan

bp

67-69 °C/11 mmHg (lit.)

mp

5-8 °C (lit.)

density

3.325 g/mL at 25 °C (lit.)

SMILES string

ICI

InChI

1S/CH2I2/c2-1-3/h1H2

InChI key

NZZFYRREKKOMAT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qingyu Kong et al.
The journal of physical chemistry. A, 109(45), 10451-10458 (2006-07-13)
Various molecular species are known to form during the photoreaction of C2H4I2 in the gas phase and in solution. We have studied all species involved in this reaction by ab initio and density functional theory (DFT) calculations: Geometries, energies, and
Eun Chul Cho et al.
Langmuir : the ACS journal of surfaces and colloids, 24(18), 9974-9978 (2008-08-21)
This study presents a method to measure the contact angles of oils on a substrate in water. Diiodomethane and perfluorodecalin were used as model oils. Self-assembled monolayers (SAMs) were prepared by adjusting the mole ratio of CH 3- and OH-terminated
Shaoliang Zhang et al.
Physical chemistry chemical physics : PCCP, 13(24), 11671-11677 (2011-05-20)
Flash photolysis (FP) coupled to resonance fluorescence (RF) was used to measure the absolute rate coefficients (k(1)) for the reaction of OH(X(2)Π) radicals with diiodomethane (CH(2)I(2)) over the temperature range 295-374 K. The experiments involved time-resolved RF detection of the
Vasudha Sharma et al.
Journal of combinatorial chemistry, 12(3), 327-331 (2010-03-12)
A mild and efficient protocol for the synthesis of a 2,6-disubstituted pyrimidine-5-carboxylate library via a Morita-Baylis-Hillman (MBH) adduct is described. Herein, the three step methodology involves the use of substituted alpha-iodomethylene beta-keto ester intermediates obtained after oxidation of the MBH
Jeongho Kim et al.
The journal of physical chemistry. A, 116(11), 2713-2722 (2011-12-01)
We investigate the structural dynamics of iodine elimination reaction of 1,2-diiodoethane (C(2)H(4)I(2)) in cyclohexane by applying time-resolved X-ray liquidography (TRXL). The TRXL technique combines structural sensitivity of X-ray diffraction and 100 ps time resolution of X-ray pulses from synchrotron and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service