Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Diiodomethane
Synonym(s):
Diiodomethane, Methylene iodide
Empirical Formula (Hill Notation):
CH
2
I
2
CAS Number:
75-11-6
Molecular Weight:
267.84
EC Number:
200-841-5
Products
Brand
Sigma-Aldrich (3)
Supelco (1)
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
8.18153
Diiodomethane,
(stabilised) for synthesis
Expand
View Pricing
158429
Diiodomethane,
ReagentPlus
®
, 99%, contains copper as stabilizer
Expand
View Pricing
1.06053
Diiodomethane,
for the separation of minerals
Expand
View Pricing
07-3750
Diiodomethane,
SAJ first grade, ≥98.0%
Expand
View Pricing