Skip to Content
Merck
All Photos(1)

Key Documents

S8636

Sigma-Aldrich

Sodium pyruvate solution

100 mM, sterile-filtered, BioReagent, suitable for cell culture

Synonym(s):

Α-ketopropionic acid sodium salt, 2-oxopropanoic acid sodium salt, Pyruvic acid sodium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCOONa
CAS Number:
Molecular Weight:
110.04
MDL number:
UNSPSC Code:
12161902
PubChem Substance ID:
NACRES:
NA.75
Pricing and availability is not currently available.

sterility

sterile-filtered

Quality Level

product line

BioReagent

form

solution

concentration

100 mM

technique(s)

cell culture | mammalian: suitable
single cell analysis: suitable

impurities

endotoxin, tested

storage temp.

2-8°C

SMILES string

[Na+].CC(=O)C([O-])=O

InChI

1S/C3H4O3.Na/c1-2(4)3(5)6;/h1H3,(H,5,6);/q;+1/p-1

InChI key

DAEPDZWVDSPTHF-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Sodium pyruvate is used by cells as an easily accessible carbohydrate source. Additionally, it is involved with amino acid metabolism and initiates the Kreb′s cycle. The 100 mM solution should be diluted 1:100 for most cell culture applications.
Sodium pyruvate solution has been used in different type of cell culture media.[1][2][3]

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dashan Dong et al.
Light, science & applications, 9, 11-11 (2020-02-07)
The emergence of super-resolution (SR) fluorescence microscopy has rejuvenated the search for new cellular sub-structures. However, SR fluorescence microscopy achieves high contrast at the expense of a holistic view of the interacting partners and surrounding environment. Thus, we developed SR
Deyanira Pérez-Morales et al.
PLoS pathogens, 13(7), e1006497-e1006497 (2017-07-14)
The evolution of bacterial pathogenicity, heavily influenced by horizontal gene transfer, provides new virulence factors and regulatory connections that alter bacterial phenotypes. Salmonella pathogenicity islands 1 and 2 (SPI-1 and SPI-2) are chromosomal regions that were acquired at different evolutionary
Refael Minnes et al.
Scientific reports, 7(1), 4381-4381 (2017-07-01)
The vast majority of cancer related deaths are caused by metastatic tumors. Therefore, identifying the metastatic potential of cancer cells is of great importance both for prognosis and for determining the correct treatment. Infrared (IR) spectroscopy of biological cells is
Eva D Papadimitraki et al.
Arthritis and rheumatism, 54(11), 3601-3611 (2006-11-01)
Toll-like receptors (TLRs) are pattern-associated receptors in innate immunity that may be involved in the recognition of self antigens and the production of pathogenic autoantibodies. This study was undertaken to examine the expression and function of various TLRs in subpopulations
Tao Zheng et al.
Journal of ethnopharmacology, 193, 691-699 (2016-10-16)
The seed of Entada phaseoloides (L.) Merr. (Entada phaseoloides) has been long used as a folk medicine for the treatment of Diabetes mellitus by Chinese ethnic minorities. Recent reports have demonstrated that total saponins from Entada phaseoloides (TSEP) could reduce

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service