Skip to Content
Merck
All Photos(1)

Key Documents

05-3040

Sigma-Aldrich

Chlorobenzene

SAJ special grade, ≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5Cl
CAS Number:
Molecular Weight:
112.56
Beilstein:
605632
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

grade

SAJ special grade

vapor density

3.86 (vs air)

vapor pressure

11.8 mmHg ( 25 °C)

Assay

≥99.5%

form

liquid

autoignition temp.

1178 °F

expl. lim.

7.1 %

availability

available only in Japan

refractive index

n20/D 1.524 (lit.)

pH

7

bp

132 °C (lit.)

mp

−45 °C (lit.)

density

1.106 g/mL at 25 °C (lit.)

SMILES string

Clc1ccccc1

InChI

1S/C6H5Cl/c7-6-4-2-1-3-5-6/h1-5H

InChI key

MVPPADPHJFYWMZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

80.6 °F

Flash Point(C)

27 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kyeongil Hwang et al.
Advanced materials (Deerfield Beach, Fla.), 27(7), 1241-1247 (2015-01-13)
Fully printed perovskite solar cells are demonstrated with slot-die coating, a scalable printing method. A sequential slot-die coating process is developed to produce efficient perovskite solar cells and to be used in a large-scale roll-to-roll printing process. All layers excluding
Zhongbing Chen et al.
Chemosphere, 89(6), 724-731 (2012-07-27)
In order to characterize the effect of vegetation on performance of constructed wetlands (CWs) treating low and high chlorinated hydrocarbon, two pilot-scale horizontal subsurface flow (HSSF) CWs (planted with Phragmites australis and unplanted) treating sulphate rich groundwater contaminated with MCB
Daniel Giesen et al.
Environmental toxicology and chemistry, 31(5), 1136-1142 (2012-03-21)
To meet the goals of Registration, Evaluation, Authorisation, and Restriction of Chemicals (REACH) as formulated by the European Commission, fast and resource-effective tools are needed to predict the toxicity of compounds in the environment. We developed quantitative structure-activity relationships (QSARs)
Wei-Chao Chen et al.
ACS applied materials & interfaces, 7(8), 4616-4622 (2015-02-11)
Controlled dispersion of single-walled carbon nanotubes (SWCNTs) in common solvents is a challenging issue, especially for the rising need of low cost flexible transparent conducting films (TCFs). Utilizing conductive polymer as surfactant to facilitate SWCNTs solubility is the most successful
Mi Yan et al.
Waste management (New York, N.Y.), 32(7), 1453-1458 (2012-04-06)
Nitrogen containing compounds such as ammonia, urea and amines can effectively inhibit the formation of polychlorinated dibenzo-p-dioxins and dibenzofurans (PCDD/Fs). Sewage sludge accumulates both sulfur and nitrogen during wastewater treatment so it could be used to reduce PCDD/Fs formation. Indeed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service