Merck
  • Side group of poly(3-alkylthiophene)s controlled dispersion of single-walled carbon nanotubes for transparent conducting film.
Side group of poly(3-alkylthiophene)s controlled dispersion of single-walled carbon nanotubes for transparent conducting film.
ACS applied materials & interfaces (2015-02-11)
Wei-Chao Chen, Hsiang-Ting Lien, Tzu-Wei Cheng, Chaochin Su, Cheong-Wei Chong, Abhijit Ganguly, Kuei-Hsien Chen, Li-Chyong Chen
ABSTRACT
Controlled dispersion of single-walled carbon nanotubes (SWCNTs) in common solvents is a challenging issue, especially for the rising need of low cost flexible transparent conducting films (TCFs). Utilizing conductive polymer as surfactant to facilitate SWCNTs solubility is the most successful pragmatic approach to such problem. Here, we show that dispersion of SWCNT with polymer significantly relies on the length of polymer side groups, which not only influences the diameter distribution of SWCNTs in solution, also eventually affects their effective TCF performance. Surfactants with longer side groups covering larger nanotube surface area could induce adequate steric effect to stabilize the wrapped SWCNTs against the nonspecific aggregation, as discerned by the optical and microscopic measurements, also evidenced from the resultant higher electrokinetic potential. This approach demonstrates a facile route to fabricate large-area SWCNTs-TCFs exhibiting high transmittance and high conductivity, with considerable uniformity over 10 cm × 10 cm.
Product Description
Supelco
Chlorobenzene, analytical standard
Sigma-Aldrich
Chlorobenzene, SAJ special grade, ≥99.5%
Sigma-Aldrich
Chlorobenzene, SAJ first grade, ≥99.0%
Sigma-Aldrich
Chlorobenzene, ReagentPlus®, 99%
Sigma-Aldrich
Chlorobenzene, ACS reagent, ≥99.5%
Sigma-Aldrich
Chlorobenzene, anhydrous, 99.8%
Sigma-Aldrich
Chlorobenzene, suitable for HPLC, 99.9%