Skip to Content
Merck
All Photos(1)

Key Documents

01-5290

Sigma-Aldrich

1-Pentanol

SAJ special grade, ≥99.0%

Synonym(s):

n-Amyl alcohol, Pentyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4OH
CAS Number:
Molecular Weight:
88.15
Beilstein:
1730975
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

grade

SAJ special grade

vapor density

3 (vs air)

vapor pressure

1 mmHg ( 13.6 °C)
1.5 mmHg ( 20 °C)

Assay

≥99.0%

form

liquid

expl. lim.

10 %, 100 °F

availability

available only in Japan

refractive index

n20/D 1.409 (lit.)

pH

7

bp

136-138 °C (lit.)

mp

−78 °C (lit.)

density

0.811 g/mL at 25 °C (lit.)

SMILES string

CCCCCO

InChI

1S/C5H12O/c1-2-3-4-5-6/h6H,2-5H2,1H3

InChI key

AMQJEAYHLZJPGS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

120.2 °F - closed cup

Flash Point(C)

49 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anthony F Cann et al.
Applied microbiology and biotechnology, 85(4), 893-899 (2009-10-28)
Pentanol isomers such as 2-methyl-1-butanol and 3-methyl-1-butanol are a useful class of chemicals with a potential application as biofuels. They are found as natural by-products of microbial fermentations from amino acid substrates. However, the production titer and yield of the
Acid/base- and anion-controllable organogels formed from a urea-based molecular switch.
Sheng-Yao Hsueh et al.
Angewandte Chemie (International ed. in English), 49(48), 9170-9173 (2010-10-19)
Kevin R Hadley et al.
The journal of physical chemistry. B, 114(13), 4590-4599 (2010-03-17)
Developing accurate models of water for use in computer simulations is important for the study of many chemical and biological systems, including lipid bilayer self-assembly. The large temporal and spatial scales needed to study such self-assembly have led to the
Nadav Amdursky et al.
The journal of physical chemistry. A, 115(24), 6481-6487 (2011-05-19)
Time-resolved emission techniques were employed to study the nonradiative process of thioflavin-T (ThT) in 1-propanol, 1-butanol, and 1-pentanol as a function of the hydrostatic pressure. Elevated hydrostatic pressure increases the alcohol viscosity, which in turn strongly influences the nonradiative rate
Gizelle Cristina Bedendo et al.
Journal of chromatography. A, 1217(42), 6449-6454 (2010-09-14)
A sensitive and precise analysis using hollow fiber renewal liquid membrane (HFRLM) extraction followed by high performance liquid chromatography-tandem mass spectrometry (LC-MS/MS) is described for determination of five sulfonamides in honey samples. In this procedure, the organic solvent introduced directly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service