THIFICDNARO
Roche
Transcriptor High Fidelity cDNA Synthesis Kit
sufficient for 50 reactions, sufficient for 100 reactions, sufficient for 200 reactions, suitable for RT-PCR, suitable for RT-qPCR
About This Item
Recommended Products
usage
sufficient for 100 reactions
sufficient for 200 reactions
sufficient for 50 reactions
Quality Level
feature
hotstart: no
manufacturer/tradename
Roche
packaging
pkg of 100 reactions (05091284001)
pkg of 200 reactions (05081963001)
pkg of 50 reactions (05081955001)
technique(s)
RT-PCR: suitable
RT-qPCR: suitable
input
purified RNA
detection method
probe-based
Related Categories
General description
High mutation rate is a hallmark of retrovirus replication. This originates in the mechanism of genome replication by the viral-encoded reverse transcriptase, which converts the genomic RNA of the virus to a dsDNA. During this process, reverse transcription produces frequent replication errors. One accepted explanation of this inaccuracy is the lack of RT 3′-5′ exonuclease activity. The naturally high error rate of reverse transcriptases is not optimal for many different applications.
The core component of the Transcriptor High Fidelity Synthesis cDNA Synthesis kit is the Transcriptor High Fidelity Reverse Transcriptase, a blend of a recombinant reverse transcriptase and a proofreading mediating enzyme. The synergy between both enzymes is the key to the ability of the enzyme blend to reverse transcribe RNA templates with 7-fold higher fidelity compared to other commonly used reverse transcriptases.
The Transcriptor High Fidelity Reverse Transcriptase enzyme blend efficiently reverse transcribes templates up to 14 kb. Due to the high thermostability of both enzyme components and the specially optimized buffer system, reverse transcription is possible at temperatures up to +55°C. This allows the reverse transcription of GC-rich templates with high secondary structure, without the need to include additives that may negatively influence the accuracy of the reverse transcription reaction.
The recombinant reverse transcriptase included in the enzyme blend is expressed in E. coli. The enzyme has RNA-directed DNA polymerase activity, DNA-dependent DNA polymerase activity, unwinding activity and RNase H activity that degrades RNA in RNA:DNA hybrids. The latter circumvents the need to perform an additional time-consuming RNase H incubation step after reverse transcription, reducing reaction time and costs.
The Transcriptor High Fidelity cDNA Synthesis Kit provides all reagents required for first-strand cDNA synthesis reactions. For priming, three different primer systems can be used. Two cDNA synthesis primers are provided with the kit: random hexamer primers and an anchored-oligo(dT)18 primer. The latter is designed to bind at the beginning of the poly(A) tail to generate full-length cDNAs and to prevent priming from internal sites of the poly(A) tail. The 5′ ends of long mRNAs are often underrepresented, therefore this priming method is preferred for most applications. The use of random hexamer primers enables priming throughout the length of RNA for uniform representation of all RNA sequences and allows reverse transcription of RNAs that do not carry a poly(A) tail. The thermostable Protector RNase Inhibitor (included in the kit) protects RNA from degradation at high reaction temperatures.
Application
- Cloning genes of interest[1]
- Sequencing transcriptomes
- Generating cDNA libraries with large and full-length inserts[2]
- Generating gene expression analysis, like RNA splicing analysis[3]
As the kit is also tested with the LightCycler® Instruments and other real-time PCR instruments, the product is also ideal for quantitative RT-PCR applications that require high fidelity.
The kit contains all components required for synthesizing cDNA suitable for direct use in qualitative RT-PCR with conventional thermal cyclers or quantitative RT-PCR on real-time PCR instruments. The 50-reaction pack size also includes 10 control reactions (control RNA and control primer mix).
Features and Benefits
- Benefit from an optimized enzyme blend with 7-fold higher fidelity compared to other commonly used reverse transcriptases.
- Rely on true-fidelity data, as determined by the sequencing of several million bases with the Genome Sequencer 20 System.
Combine accuracy with high sensitivity, high yield, and full-length transcripts.
- Generate full-length transcripts up to 14 kb with the anchored-oligo (dT)18 primers included in the kit .
- Obtain excellent yields.
- Detect low-copy number templates; reverse transcribe from as little as 10 pg template RNA.
- Transcribe a variety of templates, even the most difficult ones (e.g., GC-rich RNAs with high secondary structure) at temperatures up to +55°C.
Get results faster.
- Reduce reverse transcription reaction time to as little as 10 minutes.
- Be first to finish
Components
- Transcriptor High Fidelity Reverse Transcriptase (blend of a recombinant reverse transcriptase and a proofreading mediating enzyme)
- Transcriptor High Fidelity Reaction Buffer, 5x concentrated
- Protector RNase Inhibitor
- dNTP mix, PCR Grade
- Anchored-oligo (dT)18Primer
- Random Hexamer Primer
- DTT
- Water, PCR Grade
- Control RNA – total RNA fraction purified from the immortalized cell line K-562*
- Control PBGD primer mix (forward and reverse primer)*
* Included only in the 50-reaction pack size.
Quality
Other Notes
Error rate was determined by sequencing using the Genome Sequencer 20 System. RNA was reverse transcribed with the Transcriptor High Fidelity Reverse Transcriptase and a commonly used M-MuLV reverse transcriptase. After purification of the cDNA and amplification with a proofreading polymerase, the error rate of the reverse transcriptases was calculated by subtracting the error rate of the PCR control performed with plasmid DNA carrying the same sequence. The error rate of the Transcriptor High Fidelity ReverseTranscriptase is a mean value of four independent experiments in which at least 3.1 x 106 bases were sequenced. For the M-MuLV reverse transcriptase, 4.5 x 106 bases were sequenced.
Accuracy is represented as error rate -1, the average number of bases that are successfully reverse transcribed before a single transcription error is made. Transcriptor High Fidelity Reverse Transcriptase shows higher accuracy than a standard M-MuLV reverse transcriptase, resulting in lower error rate during cDNA synthesis.
Total RNA (1 μg from human muscle total RNA for the 2.3 kb fragment; 1 μg from HeLa total RNA for the 5.4 kb and 9.4 kb fragments; 2 μg of rat brain total RNA for the 12.4 kb fragment) was reverse transcribed with different reverse transcriptases, according to the manufacturers′ recommendations. A 5 μl aliquot of each cDNA reaction was subsequently amplified with Roche′s Expand Long Range dNTPack. Results show that the Transcriptor High Fidelity cDNA Synthesis Kit efficiently transcribes a broad range of fragment sizes with greater yield and specificity compared to the reverse transcriptases from other suppliers.
One microgram of HeLa total RNA was reverse transcribed at +50°C for the times indicated. Five-microliter aliquots of the cDNA reactions were amplified with primers for an 8.5 kb fragment, using Roche′s Expand Long Range dNTPack. All reactions were performed in duplicate. Results show that the Transcriptor High Fidelity cDNA Synthesis Kit efficiently transcribes RNA with high speed and accuracy.
1 μg of HeLa total RNA was reverse transcribed at +50°C for the times indicated. 5 μl aliquots of the cDNA reactions were amplified with primers for an 8.5 kb fragment, using Roche′s Expand Long Range dNTPack. All reactions were performed in duplicate
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
does not flash
Flash Point(C)
does not flash
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
PCR master mix simplifies PCR/RT-PCR with components like DNA polymerase, dNTPs, MgCl2, and buffer, available commercially or DIY.
PCR master mix simplifies PCR/RT-PCR with components like DNA polymerase, dNTPs, MgCl2, and buffer, available commercially or DIY.
PCR master mix simplifies PCR/RT-PCR with components like DNA polymerase, dNTPs, MgCl2, and buffer, available commercially or DIY.
PCR master mix simplifies PCR/RT-PCR with components like DNA polymerase, dNTPs, MgCl2, and buffer, available commercially or DIY.
Related Content
RT-qPCR detects specific targets with applications in gene expression and pathogen detection.
RT-qPCR detects specific targets with applications in gene expression and pathogen detection.
RT-qPCR detects specific targets with applications in gene expression and pathogen detection.
RT-qPCR detects specific targets with applications in gene expression and pathogen detection.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service