Skip to Content
Merck
All Photos(1)

Key Documents

93289

Sigma-Aldrich

TRI Reagent®

for DNA, RNA and protein isolation

Synonym(s):

DNA/RNA/protein extraction reagent, DNA/RNA/protein isolation reagent, DNA/RNA/protein purification reagent, single step RNA extraction reagent, total RNA extraction reagent, TRI Reagent® RNA Isolation Reagent

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
NACRES:
NA.52

grade

for DNA, RNA and protein isolation

form

liquid

usage

 mL sufficient for 107 cells
 mL sufficient for 100 mg tissue

impurities

phenol

Looking for similar products? Visit Product Comparison Guide

General description

TRI reagent® also known as TRIzol, is a monophasic solution of phenol and guanidinium isothiocyanate. It has a potential, to simultaneously solubilize biological material and denatures protein. It is widely used for deproteinizing RNA. This improved version of the single-step total RNA isolation reagent developed by Chomczynski allows a quick, economical, and efficient isolation of total RNA from different biological samples. This product is pure (A260/A280 ~1.8) and can be directly used in Northern blotting. Simultaneous isolation of DNA, RNA and proteins is also possible. 1 mL is sufficient for the extraction of 100 mg tissue or 107 cells.

Application

TRI Reagent® has been used for isolating RNA from various types of cells or tissues. It may also be used for processing tissues, cells cultured in monolayer, or cell pellets.
TRI Reagent is an improved version of the single-step total RNA isolation reagent developed by Chomczynski. The RNA isolation method based on this reagent is widely used and proven for RNA applications. It is ideal for quick, economical, and efficient isolation of total RNA or the simultaneous isolation of RNA, DNA, and proteins from samples of human, animal, plant, yeast, bacterial, and viral origin.

Features and Benefits

  • Easily scalable RNA isolation
  • Works with many sources: human, plant, yeast, bacterial, or viral
  • Better yields than traditional guanidine thiocyanate/cesium chloride methods

Legal Information

TRI Reagent is a registered trademark of Molecular Research Center, Inc.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - STOT RE 2

Target Organs

Nervous system,Kidney,Liver,Skin

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Expression of adenosine receptors and vegf during angiogenesis and its inhibition by pentoxifylline?a study using zebrafish model
Nathan J, et al.
Biomedicine and Pharmacotherapy, 84(6), 1406-1418 (2016)
Hybrid amniotic membrane dressing with green silver nanoparticles as bioengineered skin for wounds and burns: a pilot studies
Ramesh B, et al.
Journal of biotechnology and biomaterials, 7(272), 2-2 (2017)
Ruochan Chen et al.
Science advances, 5(5), eaav5562-eaav5562 (2019-05-28)
The ability of cytosolic lipopolysaccharide (LPS) to activate caspase-11-dependent nonclassical inflammasome is intricately controlled to avoid excessive inflammatory responses. However, very little is known about the regulatory role of various metabolic pathways in the control of caspase-11 activation. Here, we
Hao Zhang et al.
Molecular medicine reports, 22(2), 915-925 (2020-07-07)
Dilated cardiomyopathy (DCM) is a primary cardiomyopathy with high mortality. The aim of the present study was to identify the related genes in DCM. The four expression profiles (GSE17800, GSE21610, GSE42955 and GSE79962) downloaded from the Gene Expression Omnibus (GEO)
Purification of RNA using TRIzol (TRI reagent)
Rio D, et al.
Cold Spring Harbor Protocols, 2010(6), pdb-prot5439 (2010)

Articles

Simple DNA/RNA purification methods aid genome analysis from various sources, enhancing research efficiency.

Simple DNA/RNA purification methods aid genome analysis from various sources, enhancing research efficiency.

Simple DNA/RNA purification methods aid genome analysis from various sources, enhancing research efficiency.

Simple DNA/RNA purification methods aid genome analysis from various sources, enhancing research efficiency.

Protocols

TRI Reagent enables simultaneous DNA, RNA, and protein isolation with sample prep guidelines and troubleshooting.

TRI Reagent enables simultaneous DNA, RNA, and protein isolation with sample prep guidelines and troubleshooting.

TRI Reagent enables simultaneous DNA, RNA, and protein isolation with sample prep guidelines and troubleshooting.

TRI Reagent enables simultaneous DNA, RNA, and protein isolation with sample prep guidelines and troubleshooting.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service